HS15909
Lab Spatula
Flat end and spoon end, nickel-stainless steel (plated), L 225 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122403
NACRES:
NB.71
Recommended Products
material
nickel-stainless steel (plated)
L
225 mm
flat end L
50 mm
spoon end L × W
38 × 15
Related Categories
General description
Transfer, lift, and handle powders and granular materials with this nickel-plated, stainless steel wire glazed spatula. It has a Hayman-type end awith dimensions L 225 mm x W 225 mm.
Features and Benefits
- Made from nickel-plated, stainless steel wire
- Hayman-type end
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service