MilliporeSigma
All Photos(5)
Key Documents
HS120836
Micro Weigh Boats
white polystyrene
About This Item
UNSPSC Code:
41121812
NACRES:
NB.35
material
white polystyrene
packaging
pkg of 250 ea
manufacturer/tradename
Heathrow Scientific 120836
L × W × H
25 mm × 8.9 mm × 7.5 mm
General description
Ideal for microbalance weighing and/or distribution of very small amounts of sampling material. They are made out of anti-static polystyrene (food grade compliant) and have a holding capacity of approximately 500 μL.
Features and Benefits
  • Easy to Handle
  • Large tabs make boats easy to use, position and hold, especially with forceps
  • Superb Sample Recovery
  • Inert material enables the boat and contents to be added to solutions to aid 100% sample recovery, where protocols allow
  • Anti-static properties prevent sample retention from static
Choose from one of the most recent versions:
Certificates of Analysis (COA)