MilliporeSigma
Skip To
HS120225
Pour Boat Weighing Dishes
small, white polystyrene (Anti-static), pk of 500
Synonym(s):
weigh boat, weigh dish, weighing dish, weighing pan
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
About This Item
NACRES:
NB.35
UNSPSC Code:
41121812
Material:
white polystyrene (Anti-static)
Packaging:
pk of 500
Manufacturer/tradename:
Heathrow Scientific HS120225
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
white polystyrene (Anti-static)
packaging
pk of 500
manufacturer/tradename
Heathrow Scientific HS120225
L × W × H
43 mm × 58 mm × 13 mm
General description
Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Features and Benefits
  • biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • temperature range -10 to 70C
  • disposable
  • pour-spout design easily dispenses samples without flexing the dish
  • flat-bottom dishes are made from 12 mil think polystyrene for better strength, stability, and handling
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
HS1419AHS120227HS120226
description
small, white polystyrene (Anti-static), pk of 500
description
small, white polystyrene, pack of 500 ea
description
large, white polystyrene (Anti-static), pk of 500
description
medium, white polystyrene (Anti-static), pk of 500
manufacturer/tradename
Heathrow Scientific HS120225
manufacturer/tradename
Heathrow Scientific HS1419A
manufacturer/tradename
Heathrow Scientific HS120227
manufacturer/tradename
Heathrow Scientific HS120226
material
white polystyrene (Anti-static)
material
white polystyrene
material
white polystyrene (Anti-static)
material
white polystyrene (Anti-static)
packaging
pk of 500
packaging
pack of 500 ea
packaging
pk of 500
packaging
pk of 500
L × W × H
43 mm × 58 mm × 13 mm
L × W × H
43 mm × 58 mm × 13 mm
L × W × H
108 mm × 183 mm × 26 mm
L × W × H
83 mm × 132 mm × 26 mm
Still not finding the right product?
Explore all of our products under Pour Boat Weighing Dishes
Choose from one of the most recent versions:
Certificates of Analysis (COA)