Key Documents
HS120227
Pour Boat Weighing Dishes
large, white polystyrene (Anti-static), pk of 500
Synonym(s):
weigh boat, weigh dish, weighing dish, weighing pan
About This Item
UNSPSC Code:
41121812
NACRES:
NB.35
material
white polystyrene (Anti-static)
packaging
pk of 500
manufacturer/tradename
Heathrow Scientific HS120227
L × W × H
108 mm × 183 mm × 26 mm
General description
Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Features and Benefits
  • biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • temperature range -10 to 70C
  • disposable
  • pour-spout design easily dispenses samples without flexing the dish
  • flat-bottom dishes are made from 12 mil think polystyrene for better strength, stability, and handling