Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA063275

Sigma-Aldrich

Anti-ITLN1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ20022, Anti-HL-1, Anti-ITLN, Anti-LFR, Anti-hIntL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human
citations:
4
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GWSTDEANTYFKEWTCSSSPSLPRSCK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITLN1(55600)

General description

Omentin (intelectin-1) is highly expressed in omental adipose tissue, small intestine and various other tissues. It is a plasma adipokine made up of 313 amino acids.

Immunogen

intelectin 1 (galactofuranose binding)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Omentin (intelectin-1) has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes. It increases protein kinase B (AKT) phosphorylation in the absence and presence of insulin. Omentin was suggested to play a role in the defense system against microorganisms by recognizing carbohydrate chains of pathogens and bacterial components containing galactofuranosyl residues. It has anti-inflammatory and insulin-sensitizing properties. The protein may be a biomarker of metabolic disorders. It has been shown to be over-expressed in human malignant pleural mesothelioma (MPM).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST88026

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Circulating levels of adipocytokine omentin-1 in patients with renal cell cancer.
Shen XD
Cytokine, 77, 50-55 (2016)
Association of serum omentin levels with cardiac autonomic neuropathy in patients with type 2 diabetes mellitus: a hospital-based study.
Jung CH
Cardiovascular Diabetology, 14, 140-140 (2015)
Identification of omentin as a novel depot-specific adipokine in human adipose tissue: possible role in modulating insulin action.
Yang RZ
American Journal of Physiology. Endocrinology and Metabolism, 290(6), E1253-E1261 (2006)
Intelectin 1 suppresses tumor progression and is associated with improved survival in gastric cancer.
Li D
Oncotarget, 6(18), 16168-16182 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service