Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA006593

Sigma-Aldrich

Anti-PI15 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-protease inhibitor 15 preproprotein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LLFSLLCEASTVVLLNSTDSSPPTNNFTDIEAALKAQLDSADIPKARRKRYISQNDMIAILDYHNQVRGKVFPPAANMEYMVWDENLAKSAEAWAATCIWDHGPSYLLRFLGQNLSVRTGRYRSILQLVKPWYDEVKDY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PI15(51050)

Immunogen

protease inhibitor 15 preproprotein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PI15 (peptidase inhibitor 15) is a trypsin-binding protein with a molecular weight of 25kDa. It is expressed in the brain, placenta, and lymphocytes including neuroblastoma and glioblastoma cell lines. It is a novel protein belonging to the cysteine-rich secreted protein family (CRISP). It shows weak inhibitory action against trypsin. Studies have suggested that PI15 may play a role in the central nervous system.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70817

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D M Smith et al.
Mechanisms of development, 102(1-2), 223-226 (2001-04-05)
We have isolated a new chicken gene that is a member of the cysteine-rich secreted protein family (CRISP). The CRISP family is composed of over 70 members that are found in many phyla of organisms, including: vertebrates, plants, fungi, yeast
T Yamakawa et al.
Biochimica et biophysica acta, 1395(2), 202-208 (1998-02-25)
A novel trypsin inhibitor (P25TI) with an apparent molecular size of 25 kDa has previously been purified from the culture medium of human glioblastoma cells. In this study, the cDNA encoding P25TI was isolated by the polymerase chain reaction (PCR)
N Koshikawa et al.
Journal of biochemistry, 119(2), 334-339 (1996-02-01)
Our previous studies have shown that some human cancer cell lines produce pancreatic trypsinogen, plasminogen, and tissue-type kallikrein. To understand the regulatory mechanism of these proteinases, serine proteinase inhibitors secreted by human glioblastoma cell line T98G were analyzed by gelatin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service