Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA028397

Sigma-Aldrich

Anti-USP21 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-USP16, Anti-USP23, Anti-ubiquitin specific peptidase 21

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

CEAVNPTRFRAVFQKYVPSFSGYSQQDAQEFLKLLMERLHLEINRRGRRAPPILANGPVPSPPRRGGALLEEPELSDDDRANLMWKRYLEREDSKIVDLF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USP21(27005)

General description

USP21 gene is located on the human chromosome 1q23.3.
Ubiquitin specific peptidase 21 (USP21) is a protease consisting of 381 amino acid residues and a molecular mass of 43kDa. It is part of the ubiquitin-specific protease family.

Immunogen

ubiquitin specific peptidase 21 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry and immunoprecipitation against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-USP21 antibody produced in rabbit has been used in western blotting and immunohistochemistry.

Biochem/physiol Actions

Ubiquitin specific peptidase 21 (USP21) removes ubiquitin from ubiquitinated proteins and neural precursor cell expressed, developmentally down-regulated 8 (NEDD8) from NEDD8-conjugated proteins. It may have a role in cell growth regulation and is involved in immune responses. USP21 deubiquitinates interleukin-33 and stabilizes it. The protein acts as a negative regulator of nuclear factor-kappa B pathway by deubiquitinating receptor-interacting protein 1 (RIP1).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76844

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of a novel isopeptidase with dual specificity for ubiquitin- and NEDD8-conjugated proteins.
Gong L
The Journal of Biological Chemistry, 275(19) (2000)
Markus Riester et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(7), 1873-1883 (2014-02-04)
Metastatic urothelial carcinoma of the bladder is associated with multiple somatic copy-number alterations (SCNAs). We evaluated SCNAs to identify predictors of poor survival in patients with metastatic urothelial carcinoma treated with platinum-based chemotherapy. We obtained overall survival (OS) and array
The centrosomal deubiquitylase USP21 regulates Gli1 transcriptional activity and stability
Claire Heride
Journal of Cell Science (2016)
Deubiquitination and stabilization of IL-33 by USP21
Lianquin Tao
International Journal of Clinical and Experimental Pathology, 7(8) (2014)
Hung Thanh Nguyen et al.
Oncotarget, 8(38), 64095-64105 (2017-10-04)
The Hippo pathway, which acts to repress the activity of YAP and TAZ trancriptional co-activators, serve as a barrier for oncogenic transformation. Unlike other oncoproteins, YAP and TAZ are rarely activated by mutations or amplified in cancer. However, elevated YAP/TAZ

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service