Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS1819M

Sigma-Aldrich

Anti-OTUD3 Antibody, clone 12A6.1

clone 12A6.1, from mouse

Synonym(s):

OTU domain-containing protein 3, EC 3.4.19.12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

12A6.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... OTUD3(23252)

Related Categories

General description

OTU domain-containing protein 3 (UniProt: Q5T2D3; also known as EC: 3.4.19.12, OTUD3) is encoded by the OTUD3 (also known as KIAA0459) gene (Gene ID: 23252) in human. OTUD3 is a cytoplasmic deubiquitinating enzyme that hydrolyzes Lys-6- and Lys-11-linked polyubiquitin and is reported to hydrolyze heterotypic (mixed and branched) and homotypic chains. It has an OUT domain (aa 65-189) and a UBA-like domain (aa 230-270). Its specificity is given by the S1′ ubiquitin-binding site within the OTU domain composed of the Cys-, His- and Variable-loops. The UBA-like domain has no effect on ubiquitin hydrolysis. It displays a thiol-dependent ubiquitin-specific protease activity. It is reported to regulate PTEN stability and suppresses tumorigenesis. OTUD3 transgenic mice exhibit higher levels of the PTEN and are shown to be less prone to tumorigenesis. OTUD3 is believed to regulate PTEN in a manner, which is dependent on its DUB activity. The catalytically inactive mutant C76A does not upregulate PTEN and may even have a downregulating effect. It can remove Lys-6, Lys-11, Lys-27, and Lys-48 types of ubiquitin chain on PTEN. (Ref.: Yuan, L., et al (2015). Nat. Cell Biol. 17(9); 1169-1181).

Specificity

Clone 12A6.1 detects OTU domain-containing protein 3 in human cells. It iargets an epitope within 115 amino acids from the C-terminal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 115 amino acids from the C-terminal half of human OTU domain-containing protein 3.

Application

Anti-OTUD3, clone 12A6.1, Cat. No. MABS1819, is a mouse monoclonal antibody that detects OTUD3 in human cells and has been tested for use in Western Blotting.
Research Category
Signaling

Quality

Evaluated by Western Blotting in human cerebral cortex tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected OTUD3 in 10 µg of human cerebral cortex tissue lysate.

Target description

~48 kDa observed; 45.12 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meng Wang et al.
Nature communications, 12(1), 7006-7006 (2021-12-03)
Nicotine addiction and the occurrence of lymph node spread are two major significant factors associated with esophageal cancer's poor prognosis; however, nicotine's role in inducing lymphatic metastasis of esophageal cancer remains unclear. Here we show that OTU domain-containing protein 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service