Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA019885

Sigma-Aldrich

Anti-IL17RC antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-IL-17 receptor C, Anti-IL-17RC, Anti-IL-17RL, Anti-IL17Rhom, Anti-Interleukin-17 receptor C, Anti-Interleukin-17 receptor homolog, Anti-Interleukin-17 receptor-like protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

WDSDILCLPGDIVPAPGPVLAPTHLQTELVLRCQKETDCDLCLRVAVHLAVHGHWEEPEDEEKFGGAADSGVEEPRNASLQAQVVLSFQAYPTARCVLLEVQVPAALVQFGQSVGSVVYDCFEAALGSEVRIWSYTQPRYEKELNH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL17RC(84818)

General description

IL17RC (Interleukin 17 receptor C) is a novel single-pass transmembrane protein belonging to the interleukin-17 cytokine family. It is expressed in T cells and B cells with an extracellular N terminal domain. It is widely distributed in prostate, cartilage, kidney, liver, heart, and muscle, whereas at a lower level in thymus and leukocytes. It is mapped on human chromosome 3p25.3-3p24.1. Its expression has also been found in various cancerous cells such as prostate cancer.

Immunogen

Interleukin-17 receptor C Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

IL17RC (Interleukin 17 receptor C) is involved in the cytokine signaling pathway as a decoy receptor. It is essentially required for the signaling of IL-17A, IL-17F, and IL-17A/F both in vitro and in vivo. In nonhemopoietic tissues, it couples with IL-17RA and forms a complex, which drives the functions of IL-17A and IL-17F homodimers as well as IL-17A/F heterodimers. Inherited deficiency of IL17RC causes an autosomal recessive disorder, chronic mucocutaneous candidiasis (CMC), with recurrent or persistent infections of the skin, nail, oral, and genital mucosae.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72212

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kosuke Noma et al.
Journal of clinical immunology, 44(1), 18-18 (2023-12-22)
Inborn errors of the IL-17A/F-responsive pathway lead to chronic mucocutaneous candidiasis (CMC) as a predominant clinical phenotype, without other significant clinical manifestations apart from mucocutaneous staphylococcal diseases. Among inborn errors affecting IL-17-dependent immunity, autosomal recessive (AR) IL-17RC deficiency is a
Yan Hu et al.
Journal of immunology (Baltimore, Md. : 1950), 184(8), 4307-4316 (2010-03-17)
It has been suggested that IL-17RC forms a complex with IL-17RA to mediate the functions of IL-17A and IL-17F homodimers as well as IL-17A/F heterodimers. It is still unclear whether IL-17RC is absolutely required for the signaling of IL-17 cytokines
Dominik Haudenschild et al.
The Journal of biological chemistry, 277(6), 4309-4316 (2001-11-14)
Members of the interleukin-17 cytokine family are present in a variety of tissues (1-3), although the founding member, interleukin-17, is expressed exclusively in T cells and B cells (4-8). The cloning and characterization of a novel single-pass transmembrane protein with
Daniela Knizkova et al.
Nature immunology, 23(11), 1644-1652 (2022-10-22)
Interleukin-17A (IL-17A) is a key mediator of protective immunity to yeast and bacterial infections but also drives the pathogenesis of several autoimmune diseases, such as psoriasis or psoriatic arthritis. Here we show that the tetra-transmembrane protein CMTM4 is a subunit
Yun Ling et al.
The Journal of experimental medicine, 212(5), 619-631 (2015-04-29)
Chronic mucocutaneous candidiasis (CMC) is characterized by recurrent or persistent infections of the skin, nail, oral, and genital mucosae with Candida species, mainly C. albicans. Autosomal-recessive (AR) IL-17RA and ACT1 deficiencies and autosomal-dominant IL-17F deficiency, each reported in a single

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service