Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA019136

Sigma-Aldrich

Anti-NUDCD3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-NudC domain-containing protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000
western blot: 0.04-0.4 μg/mL

immunogen sequence

RRKEEEEAKTVSAAAAEKEPVPVPVQEIEIDSTTELDGHQEVEKVQPPGPVKEMAHGSQEAEAPGAVAGAAE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NUDCD3(23386)

General description

The gene NUDCD3 (NudC domain-containing gene 3) is mapped to human chromosome 7p13-p12. The protein localizes to the centrosome and midbody.

Immunogen

NudC domain-containing protein 3 recombinant protein epitope signature tag (PrEST)

Application

Anti-NUDCD3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

NUDCD3 (NudC domain-containing gene 3) is involved in cytokinesis by stabilization of dynein intermediate chain. Absence of NUDCD3 induces mitotic defects. Disturbances in NUDCD3 levels cause mislocalization of the dynein complex from kinetochores, spindle microtubules, and spindle poles, aggregation of dynein intermediate chain in the cytoplasm and degradation of dynein intermediate chain.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74853

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koji Okabayashi et al.
Cancer science, 103(9), 1617-1624 (2012-06-09)
Esophageal squamous cell cancer (ESCC) is one of the most common lethal tumors in the world, and development of new diagnostic and therapeutic methods is needed. In this study, cancer-testis antigen, BORIS, was isolated by functional cDNA expression cloning using
Inhibition of cytokinesis by overexpression of NudCL that is localized to the centrosome and midbody.
Yuqi Cai et al.
Cell research, 19(11), 1305-1308 (2009-10-07)
Tianhua Zhou et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(24), 9039-9044 (2006-06-07)
Cytoplasmic dynein, a minus-end-directed microtubule motor, has been implicated in many fundamental cellular processes; however, little is known regarding the underlying molecular machinery that regulates its stability. In Aspergillus nidulans, nuclear distribution gene C (nudC) has been implicated in the
Mariana Tamazato Longhi et al.
PloS one, 11(7), e0159856-e0159856 (2016-07-23)
Maspin (SerpinB5) is a non-inhibitory serpin (serine protease inhibitor) with very diverse biological activities including regulation of cell adhesion, migration, death, control of gene expression and oxidative stress response. Initially described as a tumor and metastasis suppressor, clinical data brought
David Asante et al.
Journal of cell science, 127(Pt 21), 4774-4787 (2014-09-11)
Cytoplasmic dynein-2 is the motor for retrograde intraflagellar transport (IFT), and mutations in dynein-2 are known to cause skeletal ciliopathies. Here, we define for the first time the composition of the human cytoplasmic dynein-2 complex. We show that the proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service