MilliporeSigma
Subunit composition of the human cytoplasmic dynein-2 complex.
Journal of cell science (2014-09-11)
David Asante, Nicola L Stevenson, David J Stephens
ABSTRACT
Cytoplasmic dynein-2 is the motor for retrograde intraflagellar transport (IFT), and mutations in dynein-2 are known to cause skeletal ciliopathies. Here, we define for the first time the composition of the human cytoplasmic dynein-2 complex. We show that the proteins encoded by the ciliopathy genes WDR34 and WDR60 are bona fide dynein-2 intermediate chains and are both required for dynein-2 function. In addition, we identify TCTEX1D2 as a unique dynein-2 light chain that is itself required for cilia function. We define several subunits common to both dynein-1 and dynein-2, including TCTEX-1 (also known as DYNLT1) and TCTEX-3 (also known as DYNLT3), roadblock-1 (also known as DYNLRB1) and roadblock-2 (also known as DYNLRB2), and LC8-1 and LC8-2 light chains (DYNLL1 and DYNLL2, respectively). We also find that NudCD3 associates with dynein-2 as it does with dynein-1. By contrast, the common dynein-1 regulators dynactin, LIS1 (also known as PAFAH1B1) and BICD2 are not found in association with dynein-2. These data explain why mutations in either WDR34 or WDR60 cause disease, as well as identifying TCTEX1D2 as a candidate ciliopathy gene.
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-ODF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-WDR60 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Anti-NUDCD3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-Dynein Antibody, 74 kDa Intermediate chains, cytoplasmic, clone 74.1, clone 74.1, Chemicon®, from mouse