Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

HPA019132

Sigma-Aldrich

Anti-SERPINB5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Maspin, Anti-Protease inhibitor 5, Anti-Serpin B5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat

enhanced validation

independent
orthogonal RNAseq
RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

TFCMGNIDSINCKIIELPFQNKHLSMFILLPKDVEDESTGLEKIEKQLNSESLSQWTNPSTMANAKVKLSIPKFKVEKMIDPK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SERPINB5(5268)

General description

The gene SERPINB5 is mapped to human chromosome 18q21.33. The protein localizes in the cytoplasm and nucleus. SERPINB5 is generally referred as MASPIN (Mammary serine protease inhibitor).

Immunogen

Serpin B5 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SERPINB5 is a class II tumor suppressor gene. It can mediate apoptosis and suppress cell invasion. It inhibits neovascularization and decreases tumor-linked microvessels. It is down-regulated in breast cancer cells, esophageal squamous cell carcinoma, glioma cells and lung adenocarcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74610

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Meng et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 36(8), 6231-6238 (2015-03-18)
Esophageal squamous cell carcinoma (ESCC) is one of the most aggressive cancers in the world. Epidemiological survey studies have verified that the development of ESCC relates to a complex interactive process between multiple genetic susceptibilities and environmental exposure. Serpins are
Da Eun Lee et al.
PloS one, 8(11), e78136-e78136 (2013-11-14)
Quantification of cell-free fetal DNA by methylation-based DNA discrimination has been used in non-invasive prenatal testing of fetal chromosomal aneuploidy. The maspin (Serpin peptidase inhibitor, clade B (ovalbumin), member 5; SERPINB5) gene, located on chromosome 18q21.33, is hypomethylated in the
Siqi Ma et al.
Pakistan journal of pharmaceutical sciences, 28(3 Suppl), 1059-1063 (2015-06-09)
This paper tested and analyzed the expression of ATF3 (activating transcription factor), MMP-2 (matrix metalloprotease) and maspin in tissue chip of glioma and its correlation with glioma advancement. Based on immunohistochemical staining, this paper selected 100 patients with glioma and
Fulvio Lonardo et al.
Histopathology, 65(6), 757-763 (2014-07-22)
To test whether changes in the subcellular localization of maspin parallel morphological progression in pulmonary adenocarcinoma, we compared its expression between lepidic and invasive growth patterns. Applying immunohistochemistry, we compared maspin expression in lepidic and invasive growth patterns occurring in
M T Longhi et al.
Cell communication and signaling : CCS, 19(1), 86-86 (2021-08-16)
Maspin (SERPINB5) is a potential tumor suppressor gene with pleiotropic biological activities, including regulation of cell proliferation, death, adhesion, migration and gene expression. Several studies indicate that nuclear localization is essential for maspin tumor suppression activity. We have previously shown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service