MilliporeSigma
  • PI3K-AKT, JAK2-STAT3 pathways and cell-cell contact regulate maspin subcellular localization.
PI3K-AKT, JAK2-STAT3 pathways and cell-cell contact regulate maspin subcellular localization.
Cell communication and signaling : CCS (2021-08-16)
M T Longhi, L E Silva, M Pereira, M Magalhães, J Reina, F N L Vitorino, B M Gumbiner, J P C da Cunha, N Cella
ABSTRACT
Maspin (SERPINB5) is a potential tumor suppressor gene with pleiotropic biological activities, including regulation of cell proliferation, death, adhesion, migration and gene expression. Several studies indicate that nuclear localization is essential for maspin tumor suppression activity. We have previously shown that the EGFR activation leads to maspin nuclear localization in MCF-10A cells. The present study investigated which EGFR downstream signaling molecules are involved in maspin nuclear localization and explored a possible role of cell-cell contact in this process. MCF-10A cells were treated with pharmacological inhibitors against EGFR downstream pathways followed by EGF treatment. Maspin subcellular localization was determined by immunofluorescence. Proteomic and interactome analyses were conducted to identify maspin-binding proteins in EGF-treated cells only. To investigate the role of cell-cell contact these cells were either treated with chelating agents or plated on different cell densities. Maspin and E-cadherin subcellular localization was determined by immunofluorescence. We found that PI3K-Akt and JAK2-STAT3, but not MAP kinase pathway, regulate EGF-induced maspin nuclear accumulation in MCF-10A cells. We observed that maspin is predominantly nuclear in sparse cell culture, but it is redistributed to the cytoplasm in confluent cells even in the presence of EGF. Proteomic and interactome results suggest a role of maspin on post-transcriptional and translation regulation, protein folding and cell-cell adhesion. Maspin nuclear accumulation is determined by an interplay between EGFR (via PI3K-Akt and JAK2-STAT3 pathways) and cell-cell contact. Video Abstract.
Sigma-Aldrich
Anti-SERPINB5 Antibody, clone 5C6.2, clone 5C6.2, from mouse
Sigma-Aldrich
Anti-FAM161B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SERPINB5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution