Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA018897

Sigma-Aldrich

Anti-CES2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CE-2, Anti-Carboxylesterase 2, Anti-hCE-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

PVPSIVGVNNNEFGWLIPKVMRIYDTQKEMDREASQAALQKMLTLLMLPPTFGDLLREEYIGDNGDPQTLQAQFQEMMADSMFVIPALQVAHFQCSRAPVYFYEFQHQP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CES2(8824)

General description

The gene CES2 (carboxylesterase 2) is mapped to human chromosome 16q22.1. It belongs to carboxylesterases family of proteins. CES2 is mainly present in the liver and the intestine. The protein exists as a monomer.

Immunogen

Carboxylesterase 2 Precursor recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Carboxylesterases hydrolyze drugs and toxins, thereby playing crucial role in metabolism and detoxication. Carboxylesterase 2 (CES2) has been shown to hydrolyze the benzoyl group of cocaine and the acetyl groups of 4-methylumbelliferyl acetate, heroin, and 6-monoacetylmorphine. Similarly, dabigatran etexilate (DABE) is an oral prodrug which is hydrolyzed by intestinal CES2 and liver CES1 to the active thrombin inhibitor, dabigatran (DAB).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85194

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Casey Laizure et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(2), 201-206 (2013-11-12)
Dabigatran etexilate (DABE) is an oral prodrug that is rapidly converted to the active thrombin inhibitor, dabigatran (DAB), by serine esterases. The aims of the present study were to investigate the in vitro kinetics and pathway of DABE hydrolysis by
Michela Capello et al.
JCO precision oncology, 4, 426-436 (2020-11-01)
The combination chemotherapy of fluorouracil, leucovorin, irinotecan, and oxaliplatin (FOLFIRINOX) has provided clinically meaningful improvement for pancreatic ductal adenocarcinoma (PDAC). We previously uncovered a role for the serine hydrolase carboxylesterase 2 (CES2) in mediating intratumoral activation of the prodrug irinotecan
Karishma Kailass et al.
Chemical science, 10(36), 8428-8437 (2019-12-06)
Irinotecan-based therapy is a common treatment for pancreatic cancer. To elicit its anticancer activity, the drug requires first the hydrolysis action of the enzyme human carboxylesterase 2 (hCES2). It has been established that pancreatic cancer patients have various levels of
H Schwer et al.
Biochemical and biophysical research communications, 233(1), 117-120 (1997-04-07)
A full-length cDNA coding for a putative intestinal carboxylesterase (iCE) was isolated from a human small intestine cDNA library. The cDNA has an open reading frame of 559 amino acids with up to 65% homology to other carboxylesterases of different
Oliver H Wang et al.
PloS one, 11(3), e0152300-e0152300 (2016-03-31)
MAP4K5 plays an important role in regulating a range of cellular responses and is involved in Wnt signaling in hematopoietic cells. However, its functions in human malignancies have not been studied. The major objectives of this study are to examine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service