MilliporeSigma
  • Molecular cloning and characterization of a novel putative carboxylesterase, present in human intestine and liver.
Molecular cloning and characterization of a novel putative carboxylesterase, present in human intestine and liver.
Biochemical and biophysical research communications (1997-04-07)
H Schwer, T Langmann, R Daig, A Becker, C Aslanidis, G Schmitz
ABSTRACT
A full-length cDNA coding for a putative intestinal carboxylesterase (iCE) was isolated from a human small intestine cDNA library. The cDNA has an open reading frame of 559 amino acids with up to 65% homology to other carboxylesterases of different mammalian species. The deduced amino-acid sequence contains many structural features, that are highly conserved among all carboxylesterase isoenzymes, like the serine esterase active site, an ER-retention signal and one Asn-Xxx-Thr site for N-linked carbohydrate addition. Northern blot analysis revealed that the corresponding mRNA is 3.4-3.6 kb in size and is preferentially expressed in human intestine with a weak signal also in liver. Analysis of cells from the gastrointestinal tract unveiled site-specific, transcriptional regulation of iCE, with higher expression in small intestine and lower expression in colon and rectum. The high expression in small intestine is attributable to a higher expression in jejunum compared to duodenum and ileum.