Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA018799

Sigma-Aldrich

Anti-GCN1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-GCN1L, Anti-GCN1L1, Anti-KIAA0219

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

EALVTDAGEVTEAGKAYVPPRVLQEALCVISGVPGLKGDVTDTEQLAQEMLIISHHPSLVAVQSGLWPALLARMKIDPEAFITRHLDQIIPRMTTQSPLNQSSMNAMGSLSVLSPDRVLPQLISTITA

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GCN1L1(10985)

General description

The gene GCN1L1 (general control of amino-acid synthesis 1-like protein 1) is mapped to human chromosome 12q24. In Saccharomyces cerevisiae, the protein was shown to localize in the cytoplasm. Like in Saccharomyces, human GCN1L1 contains an EF3 (translation elongation factor-3)-like domain.

Immunogen

GCN1 eIF2 alpha kinase activator homolog

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Translational activator GCN1L1 (general control of amino-acid synthesis 1-like protein 1) is a translational regulator. In Saccharomyces cerevisiae GCN1L1 regulates the eukaryotic translation initiation factor 2 α-subunit kinase (GCN2) activity. GCN2 is crucial for managing gene expression in presence of nutritional deprivation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74592

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael L Nickerson et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(18), 4935-4948 (2014-09-17)
Genetic analysis of bladder cancer has revealed a number of frequently altered genes, including frequent alterations of the telomerase (TERT) gene promoter, although few altered genes have been functionally evaluated. Our objective is to characterize alterations observed by exome sequencing
Francis Robert et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(37), 13421-13426 (2014-09-10)
Enhanced protein synthesis capacity is associated with increased tumor cell survival, proliferation, and resistance to chemotherapy. Cancers like multiple myeloma (MM), which display elevated activity in key translation regulatory nodes, such as the PI3K/mammalian target of rapamycin and MYC-eukaryotic initiation
Cátia M Pereira et al.
The Journal of biological chemistry, 280(31), 28316-28323 (2005-06-07)
Translational control directed by the eukaryotic translation initiation factor 2 alpha-subunit (eIF2alpha) kinase GCN2 is important for coordinating gene expression programs in response to nutritional deprivation. The GCN2 stress response, conserved from yeast to mammals, is critical for resistance to
M J Marton et al.
Molecular and cellular biology, 17(8), 4474-4489 (1997-08-01)
In the yeast Saccharomyces cerevisiae, phosphorylation of translation initiation factor eIF2 by protein kinase GCN2 leads to increased translation of the transcriptional activator GCN4 in amino acid-starved cells. The GCN1 and GCN20 proteins are components of a protein complex required

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service