Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA018039

Sigma-Aldrich

Anti-KCNH7 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Eag-related protein 3, Anti-Ether-a-go-go-related gene potassium channel 3, Anti-Ether-a-go-go-related protein 3, Anti-HERG-3, Anti-Potassium voltage-gated channel subfamily H member 7, Anti-Voltage-gated potassium channel subunit Kv113

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:1000- 1:2500

immunogen sequence

DNCKLRRRKLSFESEGEKENSTNDPEDSADTIRHYQSSKRHFEEKKSRSSSFISSIDDEQKPLFSGIVDSSPGIGKASGLDFEETVPTSGRMHIDKRSHSCKDITDMRSWERENAHPQPEDSSPSALQRAA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNH7(90134)

General description

Potassium voltage-gated channel, Eag related subfamily H, member 7 (KCNH7) is part of the ether-á-go-go-related (ERG) family. It is expressed in those regions of the brain which are associated with mood and cognition. The gene encoding it is localized on chromosome 2.

Immunogen

Potassium voltage-gated channel subfamily H member 7 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73192

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alfonso Martínez et al.
European journal of human genetics : EJHG, 16(7), 861-864 (2008-02-21)
A recent genome-wide scan of nonsynonymous SNPs and ulterior validation in case-control and family analyses evidenced a susceptibility locus for type 1 diabetes (T1D) on chromosome 2q24.3. We aimed at testing the effect of this locus in other autoimmune diseases
Kevin A Strauss et al.
Human molecular genetics, 23(23), 6395-6406 (2014-07-06)
We conducted blinded psychiatric assessments of 26 Amish subjects (52 ± 11 years) from four families with prevalent bipolar spectrum disorder, identified 10 potentially pathogenic alleles by exome sequencing, tested association of these alleles with clinical diagnoses in the larger

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service