Key Documents
HW11089-02
Sigma-Aldrich
Lifting Bar, 8 Loop, ∅20" Body, Angled
About This Item
UNSPSC Code:
41116107
NACRES:
RA.01
General description
SAFC has qualified EZ BioPac, from ILC Dover as their preferred bulk powder package. Manufactured with Armorflex 114 film, the EzBiopac range is a fit for purpose, scalable and intuitive offer that enables the clean, safe and efficient delivery of bulk powders into bioprocess.
This is an accessory which can be used to assist with the handling of the EZ BioPac by ILC Dover filled with cell culture media.
Other Notes
Low profile lifting bar for products DP4487, DP4600, DP4537, DP4601, DP4652