Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA015648

Sigma-Aldrich

Anti-GDF5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CDMP-1, Anti-Cartilage-derived morphogenetic protein 1, Anti-GDF-5, Anti-Growth/differentiation factor 5 precursor, Anti-Radotermin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

RYVFDISALEKDGLLGAELRILRKKPSDTAKPAAPGGGRAAQLKLSSCPSGRQPASLLDVRSVPGLDGSGWEVFDIWKLFRNFKNSAQLCLELEAWERGRAVDL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GDF5(8200)

Immunogen

Growth/differentiation factor 5 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Growth/differentiation factor 5 is an extracellular signaling protein encoded by the GDF5 gene in humans and belongs to growth factors with proregions. It is synthesized as a preproprotein and plays an important role in development of bone and cartilage. It is important for skeletal development, join formation and may have some role for the homeostasis of cartilage extracellular matrix. This gene may also be involved in osteoarthritis (OA) and may have some link between canonical Wnt signaling.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72604

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Enochson et al.
Osteoarthritis and cartilage, 22(4), 566-577 (2014-02-25)
Growth differentiation factor 5 (GDF5) is important for joint formation and associated with osteoarthritis (OA). Its role for the homeostasis of cartilage extracellular matrix (ECM) is, however, unknown. The canonical Wnt signaling pathway is also implemented in OA and activation
Tino Thieme et al.
The FEBS journal, 281(21), 4866-4877 (2014-09-02)
The structure and function(s) of the very large proregions of the transforming growth factor-β structure family are known in only a few cases. The proregion of growth and differentiation factor (GDF)5 comprises 354 residues. GDF5 therefore belongs to the group
CpG methylation regulates allelic expression of GDF5 by modulating binding of SP1 and SP3 repressor proteins to the osteoarthritis susceptibility SNP rs143383.
Reynard LN, Bui C, Syddall CM, et al.
Human Genetics, 133(8), 1059-1073 (2014)
Rajesh Katare et al.
Arteriosclerosis, thrombosis, and vascular biology, 34(2), 408-418 (2013-11-10)
CTX0E03 (CTX) is a clinical-grade human neural stem cell (hNSC) line that promotes angiogenesis and neurogenesis in a preclinical model of stroke and is now under clinical development for stroke disability. We evaluated the therapeutic activity of intramuscular CTX hNSC
Katja Stange et al.
Journal of molecular biology, 426(19), 3221-3231 (2014-08-06)
Growth and differentiation factor 5 (GDF5) plays a central role in bone and cartilage development by regulating the proliferation and differentiation of chondrogenic tissue. GDF5 is synthesized as a preproprotein. The biological function of the proregion comprising 354 residues is

Articles

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service