Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA019758

Sigma-Aldrich

Anti-CEACAM5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD66e antigen, Anti-CEA, Anti-Carcinoembryonic antigen, Anti-Carcinoembryonic antigen-related cell adhesion molecule 5, Anti-Meconium antigen 100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

QELFISNITEKNSGLYTCQANNSASGHSRTTVKTITVSAELPKPSISSNNSKPVEDKDAVAFTCEPEAQN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CEACAM5(1048)

General description

CEACAM5 (Carcinoembryonic antigen-related cell adhesion molecule 5) is a cell surface glycoprotein belonging to the membrane-bound immunoglobulin superfamily (IgSF). It is expressed in human airways. Structurally, it possesses a double reciprocal antiparallel binding model with leader sequence, V Ig-like N-terminal domain, three Ig-like internal domains, denoted as A1B1, A2B2, and A3B3.

Immunogen

Carcinoembryonic antigen-related cell adhesion molecule 5 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CEACAM5 (Carcinoembryonic antigen-related cell adhesion molecule) is involved in the CEA-mediated intercellular adhesion and signaling pathways. It facilitates antiparallel CEA-CEA binding during intercellular adhesion between cells and parallel interactions between CEA molecules on the same cell. Both the interactions finally have impacts on the clustering of molecules to the cell surface. It also acts as a tumor marker in the several types of cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74624

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reyes Gonzalez-Exposito et al.
Journal for immunotherapy of cancer, 7(1), 101-101 (2019-04-16)
The T cell bispecific antibody cibisatamab (CEA-TCB) binds Carcino-Embryonic Antigen (CEA) on cancer cells and CD3 on T cells, which triggers T cell killing of cancer cell lines expressing moderate to high levels of CEA at the cell surface. Patient
M Taheri et al.
The Journal of biological chemistry, 275(35), 26935-26943 (2000-06-24)
The homophilic binding of extracellular domains of membrane-bound immunoglobulin superfamily (IgSF) molecules is often required for intercellular adhesion and signaling. Carcinoembryonic antigen (CEA), a member of the IgSF, is a widely used tumor marker that functions in vitro as a
Birgitt Gutbier et al.
American journal of physiology. Lung cellular and molecular physiology, 309(3), L250-L261 (2015-06-07)
In patients with chronic obstructive pulmonary disease (COPD), Moraxella catarrhalis infection of the lower airways is associated with chronic colonization and inflammation during stable disease and acute exacerbations. Chronic smoke exposure induces chronic inflammation and impairs mucociliary clearance, thus contributing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service