Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA014018

Sigma-Aldrich

Anti-HCRTR1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Hypocretin receptor type 1, Anti-Orexin receptor type 1, Anti-Ox1r

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

MEPSATPGAQMGVPPGSREPSPVPPDYEDEFLRYLWRDYLYPKQYEWV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HCRTR1(3061)

General description

The expression of hypocretin receptor 1 has been observed in cerebral cortex, hippocampus, dorsal raphe, and ventral tegmental area. It is most abundantly expressed in the locus coeruleus.

Immunogen

Orexin receptor type 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-HCRTR1 antibody produced in rabbit has been used in a suspension bead array approach to profile serum and plasma proteins.
Anti-HCRTR1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

The gene HCRTR1 (hypocretin receptor 1) encodes a G-protein coupled receptor called as orexin receptor 1. It serves a s a receptor for orexin-A and orexin-B, with higher affinity for orexin-A. Orexin-A and orexin-B are neuromodulatory peptides that function in several processes including sleep-wake transitioning, regulation of food intake and autonomic output. Hcrtr1 signaling has been observed to be associated with cocaine seeking and the development of anhedonia. It may also be involved in the regulation of depression-like behavior. Mutations in the Hcrtr system has been implicated in Narcolepsy, a chronic sleep disorder. Polymorphism in HCRTR1 may be associated with a higher risk of polydipsia in Schizophrenia. The receptor-ligand complex also activates p38 mitogen-/stress-activated protein kinase and mediates caspase-dependent and -independent apoptosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72745

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susumu Tanaka et al.
Sleep, 29(5), 633-638 (2006-06-16)
The impairment of hypocretin neurotransmission system is considered to play a major role in the pathophysiology of narcolepsy. It has been hypothesized that autoimmune abnormalities underlie the etiology of narcolepsy, based on the tight association with HLA-DRB1*1501/ DQB1*0602. It remains
Jochen M Schwenk et al.
Proteomics, 10(3), 532-540 (2009-12-03)
In the pursuit towards a systematic analysis of human diseases, array-based approaches within antibody proteomics offer high-throughput strategies to discover protein biomarkers in serum and plasma. To investigate the influence of sample preparation on such discovery attempts, we report on
Yuko Fukunaka et al.
Neuromolecular medicine, 9(4), 292-297 (2007-11-14)
The underlying pathophysiology of primary polydipsia in schizophrenia (SCZ) is poorly understood. Our previous study, however, suggested that this condition may have a genetic component [Shinkai et al 2003 Am J Med Genet 119B 7-12]. Orexins, also called hypocretins, play
Jochen M Schwenk et al.
Molecular & cellular proteomics : MCP, 9(11), 2497-2507 (2010-08-05)
There is a need for high throughput methods for screening patient samples in the quest for potential biomarkers for diagnostics and patient care. Here, we used a combination of undirected target selection, antibody suspension bead arrays, and heat-induced epitope retrieval
Michael M Scott et al.
Behavioural brain research, 222(2), 289-294 (2011-03-08)
The orexin/hypocretin system has the potential to significantly modulate affect, based on both the neuroanatomical projection patterns of these neurons and on the sites of orexin receptor expression. However, there is little data supporting the role of specific orexin receptors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service