Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA011088

Sigma-Aldrich

Anti-STIM1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Stromal interaction molecule 1 precursor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

TAKQALSEVTAALRERLHRWQQIEILCGFQIVNNPGIHSLVAALNIDPSWMGSTRPNPAHFIMTDDVDDMDEEIVSPLSMQSPSLQSSVRQRLTEPQHGLGSQRDLTHSDSESSLHMSDRQRVAPKPPQMSRAADEALNAMTSNGSH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STIM1(6786)

General description

STIM1 (stromal interaction molecule 1) is a transmembrane protein, which spans the membrane once. It predominantly resides in endoplasmic reticulum (ER), though some of it is found in the plasma membrane. It has a canonical EF-hand (cEF) domain, two EF-hand domains, a hidden non Ca2+ binding EF-hand domain (hEF), and a sterile α motif (SAM). This is present in the N-terminal of the protein, which is located towards the lumen of ER. This gene is localized to human chromosome 11p15.5, and encodes a protein of 90kDa molecular weight, which is ubiquitously expressed.

Immunogen

Stromal interaction molecule 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

STIM1 (stromal interaction molecule 1) is an integral part of SOCE (Store-operated Ca2+ entry), which is a Ca2+ entry gateway in non-excitable cells. Orai1, which is a Ca2+ channel present on plasma membrane, interacts with STIM1 present on ER, through SOAR/CAD domain present in the C-terminal of STIM1. Whenever Ca2+ levels fall low in ER, STIM1 changes to an extended conformation of C-terminal to expose SOAR/CAD domain. Also Ca2+ depletion leads to oligomerization of STIM1, through unfolding of the EF-SAM domain. STIM1 present on plasma membrane also regulates Ca2+ entry through opening of arachidonic acid calcium (ARC) channels. It acts as a tumor suppressor gene, and its expression is related to various malignancies. Studies show that up-regulation of this gene leads to tumor arrest in G401 rhabdoid tumor, rhabdomyosarcoma and rodent myoblast cell lines.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72180

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark Duquette et al.
Matrix biology : journal of the International Society for Matrix Biology, 37, 15-24 (2014-05-23)
The thrombospondins (TSPs) are a family of matricellular proteins that regulate cellular phenotype through interactions with a myriad of other proteins and proteoglycans. We have identified a novel interaction of the members of the TSP gene family with stromal interaction
Yosuke Tojyo et al.
Journal of pharmacological sciences, 125(4), 340-346 (2014-07-18)
Store-operated Ca(2+) entry (SOCE) is a ubiquitous Ca(2+) entry pathway in non-excitable cells. It is activated by the depletion of Ca(2+) from intracellular Ca(2+) stores, notably the endoplasmic reticulum (ER). In the past 9 years, it has been established that
Marc Fahrner et al.
The Journal of biological chemistry, 289(48), 33231-33244 (2014-10-25)
Store-operated Ca(2+) entry, essential for the adaptive immunity, is initiated by the endoplasmic reticulum (ER) Ca(2+) sensor STIM1. Ca(2+) entry occurs through the plasma membrane resident Ca(2+) channel Orai1 that directly interacts with the C-terminal STIM1 domain, named SOAR/CAD. Depletion
S S Manji et al.
Biochimica et biophysica acta, 1481(1), 147-155 (2000-09-27)
STIM1 is a novel candidate growth suppressor gene mapping to the human chromosome region 11p15.5 that is associated with several malignancies. STIM1 overexpression studies in G401 rhabdoid tumour, rhabdomyosarcoma and rodent myoblast cell lines causes growth arrest, consistent with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service