Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA009309

Sigma-Aldrich

Anti-LAMA3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-E170 antibody produced in rabbit, Anti-Epiligrin 170 kDa subunit antibody produced in rabbit, Anti-Laminin subunit α-3 precursor antibody produced in rabbit, Anti-Nicein subunit α antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

LLNRIRTWQKTHQGENNGLANSIRDSLNEYEAKLSDLRARLQEAAAQAKQANGLNQENERALGAIQRQVKEINSLQSDFTKYLTTADSSLLQTNIALQLMEKSQKEYE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAMA3(3909)

General description

LAMA3 (laminin subunit α3), along with LAMB3 (β3) and LAMC2 (γ2), forms a subunit of laminin5 protein. This gene is localized to human chromosome 18q11.2. Its C-terminal contains G-domain, which is a compact globular domain composed of subdomains G1-G5.

Immunogen

Laminin subunit α-3 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-LAMA3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

LAMA3 (laminin subunit α3) is a subunit of laminin5 protein, which plays an essential role in maintaining skin integrity. It is responsible for the contact between dermis and epidermis, and promotes the adhesion, spreading and migration of human keratinocytes. LAMA3 is linked with susceptibility to atopic dermatitis (AD), which is a chronic inflammatory skin disorder. Rarely found SNP in this gene is shown to have biological interactions characteristic of prion disease or with prion proteins. Studies in population with Punjabi ancestry show that a homozygous nonsense mutation in this gene is linked with laryngo-onycho-cutaneous syndrome, which is an autosomal recessive disorder and a subtype of junctional epidermolysis bullosa.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71201

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilknur Senyürek et al.
Journal of innate immunity, 6(4), 467-484 (2014-01-25)
Laminins play a fundamental role in basement membrane architecture and function in human skin. The C-terminal laminin G domain-like (LG) modules of laminin α chains are modified by proteolysis to generate LG1-3 and secreted LG4-5 tandem modules. In this study
Susanne Stemmler et al.
BMC dermatology, 14, 17-17 (2014-11-05)
Atopic dermatitis (AD) is a chronic inflammatory skin disorder caused by complex interaction of genetic and environmental factors. Besides mutations in the filaggrin gene, leading to impaired skin barrier function, variation in genes encoding additional skin proteins has been suggested
Sol Moe Lee et al.
BMC medical genomics, 7, 52-52 (2014-08-26)
Human prion diseases are caused by abnormal accumulation of misfolded prion protein in the brain tissue. Inherited prion diseases, including familial Creutzfeldt-Jakob disease (fCJD), are associated with mutations of the prion protein gene (PRNP). The glutamate (E)-to-lysine (K) substitution at
M Barzegar et al.
The British journal of dermatology, 169(6), 1353-1356 (2013-07-23)
Laryngo-onycho-cutaneous (LOC) syndrome is a subtype of autosomal recessive junctional epidermolysis bullosa in which there is prominent skin and mucosal granulation tissue that can lead to delayed wound healing, laryngeal obstruction and blindness. Thus far, all cases are of Punjabi
Elisabetta Kuhn et al.
The American journal of surgical pathology, 36(12), 1826-1834 (2012-08-16)
There is compelling evidence to suggest that serous tubal intraepithelial carcinoma (STIC) is the likely primary site for the development of many pelvic high-grade serous carcinomas (HGSCs). Identifying molecules that are upregulated in STIC is important not only to provide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service