Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA011081

Sigma-Aldrich

Anti-TMIGD2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Transmembrane and immunoglobulin domain-containing protein 2 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

CQVDQATAWERLRVKWTKDGAILCQPYITNGSLSLGVCGPQGRLSWQAPSHLTLQLDPVSLNHSGAYVCWAAVEIPELEEAEGNITRLFVDPDDPTQNRNRIA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TMIGD2(126259)

General description

TMIGD2 (transmembrane and immunoglobulin domain-containing protein 2) is a cell adhesion protein, which has one immunoglobulin (Ig) domain in its extracellular region, one transmembrane region and a cytoplasmic domain made of 110 amino acids which is rich in proline. It is highly expressed in endothelial and epithelial cells. This gene maps to human chromosome 19q13.3, spans 10.2kb and has 5 exons. The mRNA for this gene is found mainly in lymphoid organs, with highest expression in spleen, thymus, liver and peripheral blood lymphocyte.

Immunogen

Transmembrane and immunoglobulin domain-containing protein 2 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TMIGD2 (transmembrane and immunoglobulin domain-containing protein 2) is involved in modulating the morphology of cell, cell-cell interactions and cell migration. It increases cell-cell interaction and aggregation, along with increase in focal adhesions. It regulates angiogenesis by binding with various SH3 (SRC homology 3) domain containing proteins such as SPIN90 (SH3 protein interacting with Nck, 90kDa). SPIN90 also mediates cytoskeleton reorganization and therefore, controls cell adhesion. TMIGD2 is responsible for activation and co-stimulation of T-cells through AKT (v-Akt Murine Thymoma Viral Oncogene) pathway.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72192

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuwen Zhu et al.
Nature communications, 4, 2043-2043 (2013-06-21)
The B7/CD28 family has profound modulatory effects in immune responses and constitutes an important target for the development of novel therapeutic drugs against human diseases. Here we describe a new CD28 homologue (CD28H) that has unique functions in the regulation
Nader Rahimi et al.
Molecular biology of the cell, 23(9), 1646-1656 (2012-03-16)
Angiogenesis-the growth of new blood vessels from preexisting vessels-is an important physiological process and is considered to play a key role in tumor growth and metastasis. We identified the immunoglobulin-containing and proline-rich receptor-1 (IGPR-1, also called TMIGD2) gene as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service