Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP5820715006

Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges

FA-45-6-30, aerosol tight, to hold, 6 x 15/50 mL conical tubes, lid

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909
NACRES:
NB.82

feature

lid
to hold 6 x 15/50 mL conical tubes
aerosol tight
high-speed fixed angle rotor

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5820715006 (Global)

parameter

12,000 rpm
20,130 × g max. RCF (16,639 x g for 5810R)

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5804/5804R/5810/5810R

Looking for similar products? Visit Product Comparison Guide

General description

  • High-speed fixed angle rotor holds 6 × 15/50 mL conical tubes.
  • Features a QuickLock rotor lid for ergonomic lid locking.
  • The aerosol-tight cap facilitates safe centrifugation of potentially hazardous materials.
  • Portable, low weight for easy lifting.
  • Rotors are made of anodized aluminum for high chemical resistance and to prevent corrosion.
  • 45° borehole angle minimizes pellets smear.
  • Rotor, lid and adapters are autoclavable.
A wide selections of rotors to fit the Eppendorf 5804 and 5810 centrifuges.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service