Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA007393

Sigma-Aldrich

Anti-ANXA4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-35-β Calcimedin antibody produced in rabbit, Anti-Annexin A4 antibody produced in rabbit, Anti-Annexin IV antibody produced in rabbit, Anti-Carbohydrate-binding protein P33/P41 antibody produced in rabbit, Anti-Chromobindin-4 antibody produced in rabbit, Anti-Endonexin I antibody produced in rabbit, Anti-Lipocortin IV antibody produced in rabbit, Anti-P32.5 antibody produced in rabbit, Anti-P33/41 antibody produced in rabbit, Anti-PAP-II antibody produced in rabbit, Anti-PP4-X antibody produced in rabbit, Anti-Placental anticoagulant protein II antibody produced in rabbit, Anti-Protein II antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

KGGTVKAASGFNAMEDAQTLRKAMKGLGTDEDAIISVLAYRNTAQRQEIRTAYKSTIGRDLIDDLKSELSGNFEQVIVGMMTPTVLYDVQELRRAMKGAGTDEGCLIEILASRTPEEIRRISQTYQQQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANXA4(307)

General description

ANXA4 (annexin A4) gene encodes a protein that is a member of the annexin family of calcium-dependent phospholipid binding proteins. These proteins contain divergent NH2-terminal ′head′ domain that confers specific properties to the individual annexin and a conserved 34kDa COOH-terminal protein core that binds to calcium and membrane. Annexin 4 was first identified in homogenates of human placenta. It is expressed in several tissues such as secretory epithelia in the lung, stomach, intestine, and kidney. It is associated with polarized epithelial cells.

Immunogen

Annexin A4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ANXA4 (annexin A4) may function in the regulation of chloride ions across the epithelial membrane. It has been suggested to play a role in normal pronephrogenesis in Xenopus laevis. It has been identifies as a candidate biomarker for cisplatin resistance in malignant mesothelioma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70189

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuya Yamashita et al.
Biochemical and biophysical research communications, 421(1), 140-144 (2012-04-14)
Mesothelioma is a highly malignant tumor with a poor prognosis and limited treatment options. Although cisplatin (CDDP) is an effective anticancer drug, its response rate is only 20%. Therefore, discovery of biomarkers is desirable to distinguish the CDDP-susceptible versus resistant
Karine L Massé et al.
Organogenesis, 3(2), 83-92 (2007-10-01)
Anxa4 belongs to the multigenic annexin family of proteins which are characterized by their ability to interact with membranes in a calcium-dependent manner. Defined as a marker for polarized epithelial cells, Anxa4 is believed to be involved in many cellular
Yi-Yao Liu et al.
American journal of cancer research, 7(6), 1285-1297 (2017-07-04)
The occurrence and progression of hepatocellular carcinoma (HCC) are affected by complicated signal transduction factors. Our previous study identified Ikaros as a novel reactivated therapeutic target that acts as a transcriptional repressor and reactivates anticancer mechanisms in HCC therapy. Annexin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service