Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

370R-1

Sigma-Aldrich

LMO2 (SP51) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP51, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (370R-14)
vial of 0.5 mL concentrate (370R-15)
bottle of 1.0 mL predilute (370R-17)
vial of 1.0 mL concentrate (370R-16)
bottle of 7.0 mL predilute (370R-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:500-1:2000

isotype

IgG

control

diffuse large B-cell, follicular lymphoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... LMO2(4005)

General description

LMO2 is expressed in normal germinal center B-cells and in a subset of lymphomas derived from those cells in addition to bone marrow hematopoietic precursors and endothelial cells. LMO2 protein expression has also been shown to play an important role in the diagnosis of diffuse large B-cell lymphomas, regardless of rituximab treatment. It also plays a role in angiogenesis and hematopoiesis. It is weakly expressed in mantle zone B-cells but not in mantle cell or marginal zone lymphomas. SF Younes et al. Have demonstrated LMO2 expression in 70% of follicular lymphomas. These data suggest that anti-LMO2 is a useful adjunct in the diagnosis of follicular lymphoma (FL). As LMO2 appears not to be down regulated in higher grade FL or the interfollicular and diffuse components of FL, its utility in variant immunoarchitectural patterns of FL and in cases that lack CD10 and bcl2, is similar to that of HGAL. One advantage of LMO2 is its crisp nuclear localization that allows for easier interpretation than diffuse cytoplasmic staining pattern.

Quality


IVD

IVD

IVD

RUO

Linkage

LMO2 Positive Control Slides, Product No. 370S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheren F Younes et al.
The American journal of surgical pathology, 34(9), 1266-1276 (2010-08-11)
Follicular lymphoma (FL) can exhibit variant histologic patterns that can lead to confusion with other B-cell lymphomas and reactive conditions. Diagnostic markers such as CD10 and BCL2 may be difficult to interpret in variant FL patterns, and are often diminished

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service