Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA005474

Sigma-Aldrich

Anti-BRIP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BACH1, Anti-BRCA1 interacting protein C-terminal helicase 1, Anti-FANCJ, Anti-OF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

FNKQTKRVSWSSFNSLGQYFTGKIPKATPELGSSENSASSPPRFKTEKMESKTVLPFTDKCESSNLTVNTSFGSCPQSETIISSLKIDATLTRKNHSEHPLCSEEALDPDIELSLVSEEDKQSTSNRDFETEAEDESIYFTPEL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BRIP1(83990)

General description

BRCA1 interacting protein C-terminal helicase 1 (BRIP1) is an endogenous protein belonging to DEAH family of DNA helicases. It is an interacting partner of BRCA1 protein. It is a nuclear protein and functions both as an ATP-dependent DNA helicase and DNA-dependent ATPase. This gene is located on chromosome 17q22, is 180kb long and contains 20 exons. BRIP1 protein is composed of 1,249 amino acids and is universally expressed.

Immunogen

BRCA1 interacting protein C-terminal helicase 1 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

BRCA1 interacting protein C-terminal helicase 1 (BRIP1) plays a key role in BRCA1-dependent DNA repair and checkpoint activities, and interacts with the BRCT domain of BRCA1. It aids BRCA1 in its tumor suppressor function and the repair of DNA double strand breaks by forming a complex with it. Mutations in BRIP1 are linked with Fanconi anemia, which is characterized by predisposition to cancer, developmental abnormalities and bone marrow failure. Studies suggest that mutations in this gene are linked to early onset breast cancer. Inactivation of BRIP1 gene leads to disruption of mammary morphogenesis and causes aberrant mammary acinar morphogenesis. Studies show that polymorphisms in this gene are linked to susceptibility to cervical cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86956

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Zou et al.
Oncology letters, 11(1), 551-558 (2016-02-13)
Breast cancer 1, early onset (BRCA1)-interacting protein 1 (BRIP1), a DNA-dependent adenosine triphosphatase and DNA helicase, is required for BRCA-associated DNA damage repair functions, and may be associated with the tumorigenesis and aggressiveness of various cancers. The present study investigated
X D Ma et al.
Gene, 524(2), 208-213 (2013-05-07)
BRIP1 (BRCA1-interacting protein 1), a DNA-dependent ATPase and a DNA helicase, is critical for BRCA-associated DNA damage repair functions, and may be involved in the development of cervical cancer. Genetic markers in different regions of the BRIP1 gene have a
Kazuhiro Daino et al.
PloS one, 8(9), e74013-e74013 (2013-09-17)
BRIP1 is a DNA helicase that directly interacts with the C-terminal BRCT repeat of the breast cancer susceptibility protein BRCA1 and plays an important role in BRCA1-dependent DNA repair and DNA damage-induced checkpoint control. Recent studies implicate BRIP1 as a
Honglin Song et al.
PloS one, 2(3), e268-e268 (2007-03-08)
BRIP1 interacts with BRCA1 and functions in regulating DNA double strand break repair pathways. Germline BRIP1 mutations are associated with breast cancer and Fanconi anemia. Thus, common variants in the BRIP1 are candidates for breast and ovarian cancer susceptibility. We
Wendy L Bridge et al.
Nature genetics, 37(9), 953-957 (2005-08-24)
BRIP1 (also called BACH1) is a DEAH helicase that interacts with the BRCT domain of BRCA1 (refs. 1-6) and has an important role in BRCA1-dependent DNA repair and checkpoint functions. We cloned the chicken ortholog of BRIP1 and established a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service