Skip to Content
MilliporeSigma

B6542

Brefeldin A

≥99% (HPLC and TLC), BioXtra, Molecular Biology

Synonym(s):

γ,4-Dihydroxy-2-(6-hydroxy-1-heptenyl)-4-cyclopentanecrotonic acid λ-lactone, Ascotoxin, BFA, Cyanein, Decumbin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H24O4
CAS Number:
Molecular Weight:
280.36
UNSPSC Code:
12352200
NACRES:
NA.52
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
25191
Assay:
≥99% (HPLC and TLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Penicillium brefeldianum

Quality Segment

grade

Molecular Biology

product line

BioXtra

assay

≥99% (HPLC and TLC)

form

powder

mol wt

280.36

solubility

DMSO: 10 mg/mL (Store stock solutions at -20 °C)

antibiotic activity spectrum

neoplastics

mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@H](O)\C=C\C(=O)O1

InChI

1S/C16H24O4/c1-11-5-3-2-4-6-12-9-13(17)10-14(12)15(18)7-8-16(19)20-11/h4,6-8,11-15,17-18H,2-3,5,9-10H2,1H3/b6-4+,8-7+/t11-,12+,13-,14+,15+/m0/s1

InChI key

KQNZDYYTLMIZCT-KQPMLPITSA-N

General description

Brefeldin induces ADP-ribosylation of CtBP1/BARS protein that regulates the intracellular trafficking and movement of secretory proteins from endoplasmic reticulum to Golgi apparatus.
Chemical structure: macrolide

Application

Suitable for molecular biology studies of secretory proteins and mechanisms of intracellular transport

Biochem/physiol Actions

Brefeldin A (BFA) is a fungal metabolite which disrupts the structure and function of the Golgi apparatus. BFA is an activator of the sphingomyelin cycle. Brefeldin A-mediated apoptosis has been observed in human tumor cells.
Brefeldin is a fungal metabolite that disrupts protein secretion in eukaryotic cells by disrupting the Golgi apparatus. It does not affect the process of protein synthesis.
Disrupts the structure and function of the Golgi apparatus; activator of the sphingomyelin cycle.


Still not finding the right product?

Explore all of our products under Brefeldin A


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library