Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA004337

Sigma-Aldrich

Anti-IFITM3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Interferon-induced transmembrane protein 3, Anti-Interferon-inducible protein 1-8U

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

MNHTVQTFFSPVNSGQPPNYEMLKEEHEVAVLGAPHNPAPPTSTVIHIRSETSVPDH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFITM3(10410)

General description

IFITM3 (interferon induced transmembrane protein 3) is a transmembrane-domain protein (5-18kDa) belongs to a family of murine genes. It acts as a cellular restriction factor. It is associated with various cellular processes such as cell adhesion, immune-cell regulation, germ cell homing and maturation, and bone mineralization. It is involved in the inhibition of infection by influenza virus and many other pathogenic viruses.

Immunogen

Interferon-induced transmembrane protein 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The activity of IFITM3 (interferon induced transmembrane protein 3) is post-translationally modulated by S-palmitoylation and lysine ubiquitination. IFITM3 shows controlled clustering in membrane compartments and antiviral activity against influenza virus in post S-palmitoylated condition. It has reported that S-palmitoylation positively regulates IFITM3 membrane affinity and antiviral activity. On the contrary, ubiquitination decreases localization with endolysosomes and decreases antiviral activity. It has also corroborated that gene silencing of IFITM3 plays an important role in glioma cell growth and migration. It can be used as a tumor biomarker as it possesses characteristics of inhibiting tumor development.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85237

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Song Wang et al.
Annals of clinical and translational neurology, 8(3), 558-570 (2021-01-20)
Rasmussen's encephalitis (RE) is a rare and severe progressive epileptic syndrome with unknown etiology. Infection by viruses such as human cytomegalovirus (HCMV) has been hypothesized to be a potential trigger for RE. Interferon-induced transmembrane protein-3 (IFITM3) single-nucleotide polymorphism (SNP) rs12252
Jacob S Yount et al.
Nature chemical biology, 6(8), 610-614 (2010-07-06)
Identification of immune effectors and the post-translational modifications that control their activity is essential for dissecting mechanisms of immunity. Here we demonstrate that the antiviral activity of interferon-induced transmembrane protein 3 (IFITM3) is post-translationally regulated by S-palmitoylation. Large-scale profiling of
Bing Zhao et al.
BMC neurology, 13, 210-210 (2013-12-29)
Interferon induced transmembrane protein 3 (IFITM3) is transcribed in most tissues and highly interferon-inducible. However, the role of IFITM3 in cancer is still poorly understood. Expression levels of IFITM3 were analyzed in 60 glioma patients by immunohistochemistry (IHC). Following closely
Jacob S Yount et al.
The Journal of biological chemistry, 287(23), 19631-19641 (2012-04-19)
The interferon (IFN)-induced transmembrane protein 3 (IFITM3) is a cellular restriction factor that inhibits infection by influenza virus and many other pathogenic viruses. IFITM3 prevents endocytosed virus particles from accessing the host cytoplasm although little is known regarding its regulatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service