Skip to Content
MilliporeSigma
All Photos(6)

Documents

HPA002636

Sigma-Aldrich

Anti-AURKA antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Aurora kinase A, Anti-Aurora-A, Anti-Aurora-related kinase 1, Anti-Aurora/IPL1-related kinase 1, Anti-Breast tumor-amplified kinase, Anti-Serine/threonine kinase 15, Anti-Serine/threonine-protein kinase 6, Anti-hARK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

ISGPVKATAPVGGPKRVLVTQQFPCQNPLPVNSGQAQRVLCPSNSSQRVPLQAQKLVSSHKPVQNQKQKQLQATSVPHPVSRPLNNTQKSKQPLPSAPENNPEEELASKQKNEESKKRQWALEDFEIGRPLGKGKFGNVYL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AURKA(6790)

General description

AURKA (aurora kinase A) is a centrosome-associated serine/threonine kinase which is associated with cell cycle checkpoint. It exhibits an inductive effect on the centrosome duplication-distribution abnormalities and aneuploidy in mammalian cells.

Immunogen

Serine/threonine-protein kinase 6 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-AURKA antibody produced in rabbit has been used in immunohistochemistry and immunoblotting.
Anti-AURKA antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

All the members of aurora family kinase play a pivotal role in the execution of cell division by participating in the formation of bipolar mitotic spindle, accurate segregation of chromosomes and the completion of cytokinesis. In mammals, AURKA (aurora kinase A) overexpression shows different chromosomal abnormalities, such as centrosome amplification, chromosomal transformation and instability. It helps in increasing centrosomal γ-tubulin and two other PCM (pericentriolar material) components, ZYG-9 (zygote defective protein 9) and Ce-Grip during entry of embryos into mitosis stage. AURKA is overexpressed in breast tumor cells and may associate with the oncogenic transformation. It has been reported that AURKA has clinical relationship with non-Hodgkin′s lymphoma as it is involved not only in disease activity but also in tumorigenesis of the disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86425

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurora kinase A is a prognostic marker in colorectal adenocarcinoma
Koh HM, et al.
Journal of pathology and translational medicine, 51(1), 32-32 (2017)
Richard Crane et al.
Biology of the cell, 96(3), 215-229 (2004-06-09)
The Aurora family kinases are pivotal to the successful execution of cell division. Together they ensure the formation of a bipolar mitotic spindle, accurate segregation of chromosomes and the completion of cytokinesis. They are also attractive drug targets, being frequently
Hyun Min Koh et al.
Journal of pathology and translational medicine, 51(1), 32-39 (2016-12-26)
Aurora kinase A (AURKA), or STK15/BTAK, is a member of the serine/threonine kinase family and plays important roles in mitosis and chromosome stability. This study investigated the clinical significance of AURKA expression in colorectal cancer patients in Korea. AURKA protein
Clinicopathologic significance of Aurora kinase A expression in non-small cell lung cancer.
Lee
Basic and Applied Pathology, 5(1), 8-14 (2012)
E Hannak et al.
The Journal of cell biology, 155(7), 1109-1116 (2001-12-19)
Centrosomes mature as cells enter mitosis, accumulating gamma-tubulin and other pericentriolar material (PCM) components. This occurs concomitant with an increase in the number of centrosomally organized microtubules (MTs). Here, we use RNA-mediated interference (RNAi) to examine the role of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service