Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA001756

Sigma-Aldrich

Anti-RREB1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Finger protein in nuclear bodies, Anti-RAS-responsive element-binding protein 1, Anti-RREB-1, Anti-Raf-responsive zinc finger protein LZ321, Anti-Zep-1, Anti-Zinc finger motif-enhancer binding-protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

ATTDTNKFSPFLQTAEDNTQDEVAGAPADHHGPSDEEQGSPPEDKLLRAKRNSYTNCLQKITCPHCPRVFPWASSLQRHMLTHTGQKPFPCQKCDAFFSTKSNCERHQLRKHGVTIRRAPPLSNN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RREB1(6239)

Immunogen

RAS-responsive element-binding protein 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-RREB1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

RREB1, RAS-responsive element-binding protein 1, is a transcription factor with few splice variants such as RREB1α, RREB1β, RREB1γ, and RREB1δ. It is a 120kDa nuclear protein with ∼15 zinc finger domains, based on alternative splicing. The expression of RREB-1 is ubiquitously found in human tissues outside the adult brain. In overexpression state in thyroid cancer cell line TT, it mediates increased transactivation of the calcitonin (CT ) gene promoter reporter construct during Ras or Raf induced differentiation. It acts as a putative Ral-regulated gene.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84760

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew D Nitz et al.
The American journal of pathology, 179(1), 477-486 (2011-06-28)
RREB1 is an alternatively spliced transcription factor implicated in Ras signaling and cancer. Little is known about the expression of RREB1 isoforms in cell lines or human tumors, or about the clinical relevance of the latter. We have developed tools
A Thiagalingam et al.
Molecular and cellular biology, 16(10), 5335-5345 (1996-10-01)
An activated ras oncogene induces a program of differentiation in the human medullary thyroid cancer cell line TT. This differentiation process is accompanied by a marked increase in the transcription of the human calcitonin (CT) gene. We have localized a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service