Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABT861M

Sigma-Aldrich

Anti-CDCP1 (CD318) Antibody, clone 9A2

clone 9A2, from mouse

Synonym(s):

CUB domain-containing protein 1, Membrane glycoprotein gp140, Subtractive immunization M plus HEp3-associated 135 kDa protein, SIMA135, Transmembrane and associated with Src kinases, CD318

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9A2, monoclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CDCP1(64866)

Related Categories

General description

CUB domain-containing protein 1 (UniProt: Q9H5V8; also known as Membrane glycoprotein gp140, Subtractive immunization M plus HEp3-associated 135 kDa protein, SIMA135, Transmembrane and associated with Src kinases, CD318) is encoded by the CDCP1 (also known as TRASK) gene (Gene ID: 64866) in human. CUB domain-containing protein 1 is a single-pass membrane protein that is synthesized with a 29 amino acid signal peptide, which is subsequently cleaved to generate the mature form. A soluble form of CUB domain-containing protein 1 has also been reported that is produced by proteolytic cleavage at the cell surface. CUB domain-containing protein 1 is highly expressed in mitotic cells with low expression during interphase. It is detected at highest levels in skeletal muscle and colon. Lower expression is reported in kidney, small intestine, placenta, and lung. It is also reported in cells with phenotypes reminiscent of mesenchymal stem cells and neural stem cells. CUB domain-containing protein 1 levels are up-regulated in a number of human tumor cell lines and in colorectal cancer, breast carcinoma and lung cancer. It is reported to be involved in cell adhesion and cell matrix association and may play a role in the regulation of anchorage versus migration or proliferation versus differentiation via its phosphorylation.

Specificity

Clone 9A2 detects CUB domain-containing protein 1 in human and murine cells. It targets an epitope within the extracellular domain.

Immunogen

Epitope: extracellular domain
His-tagged recombinant fragment corresponding to 638 amino acids from the internal region of human CUB domain-containing protein 1.

Application

Anti-CDCP1 (CD318), clone 9A2, Cat. No. MABT861, is a mouse monoclonal that detects CUB domain-containing protein 1 and has been tested for use in Flow Cytometry.
Flow Cytometry Analysis: 1 µg from a representative lot detected CDCP1 CD318 in MDA-MB231 cells.


Research Category
Cell Structure

Quality

Evaluated by Flow Cytometry in DLD-1 cells.

Flow Cytometry Analysis: 1 µg of this antibody detected CDCP1 CD318 in one million DLD-1 cells.

Target description

92.93 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service