Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA001497

Sigma-Aldrich

Anti-AMBP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Alpha-1 microglycoprotein, Anti-ITI-LC, Anti-Protein HC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

VLPQEEEGSGGGQLVTEVTKKEDSCQLGYSAGPCMGMTSRYFYNGTSMACETFQYGGCMGNGNNFVTEKECLQTCRTVAACNLPIVRGPCRAFIQLWAFDAVKGKCVLFPYGGCQGNGNKFYSEKECRE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AMBP(259)

Immunogen

Protein AMBP precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

AMBP (α-1-microglobulin/bikunin precursor) gene encodes a precursor that is proteolytically cleaved into two functional proteins, α-1-microglobulin (AIM) and bikunin. The gene is localized to chromosome 9q32 – 33 region and is specifically expressed in the liver. AIM and bikunin are plasma proteins that have no relation to each other except that they are co-synthesized. A1M is a 27kDa glycoprotein that belongs to the lipocalin superfamily of hydrophobic ligand-binding proteins. It may be present both as free molecules and as complexes with either IgA heavy chains, functioning in the regulation of inflammatory processes. Bikunin is a urinary trypsin inhibitor belonging to the superfamily of Kunitz-type protease inhibitors. It forms a complex with H1, H2, and H3 heavy chains of the inter-α-inhibitor family and functions in several physiological and pathological processes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84511

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Rouet et al.
The Journal of biological chemistry, 267(29), 20765-20773 (1992-10-15)
alpha 1-Microglobulin (A1M) and bikunin are plasma proteins which are present both as free molecules and as complexes with either IgA heavy chains for A1M or the H1, H2, and H3 heavy chains of the inter-alpha-inhibitor family for bikunin. Mature
Joris Penders et al.
Clinica chimica acta; international journal of clinical chemistry, 346(2), 107-118 (2004-07-17)
Urinary microproteins are becoming increasingly important in clinical diagnostics. They can contribute in the non-invasive early detection of renal abnormalities and the differentiation of various nephrological and urological pathologies. Alpha 1-microglobulin (A1M) is an immunomodulatory protein with a broad spectrum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service