Skip to Content
MilliporeSigma
All Photos(2)

Documents

ABN40

Sigma-Aldrich

Anti-DLK1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protein delta homolog 1, DLK-1, pG2, Fetal antigen 1, FA1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DLK1(8788)

General description

Delta-like 1 homolog (DLK1), also known as fetal antigen 1 (FA1) and preadipocyte factor 1 (pref-1), is a transmembrane protein containing 6 epidermal growth factor repeats whose expression is known to modulate differentiation signals. Studies have shown DLK1 is involved in differentiation of several cell types, including adipocytes. Studies have shown deficiencies in DLK1 displaying growth retardation, obesity, skeletal malformation, and increased serum lipid. DLK1 is a paternally expressed imprinted gene that plays a role in normal development and in the maintence of homeostatsis of adipose tissue mass.

Immunogen

KLH-conjugated linear peptide corresponding to human DLK1.

Application

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected DLK1 in mouse embryonic tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
This Anti-DLK1 Antibody is validated for use in WB, IH for the detection of DLK1.

Quality

Evaluated by Western Blot in HepG2 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected DLK1 on 10 µg of HepG2 cell lysate.

Target description

~47 kDa observed. The calculated molecular weight of this protein is 41 kDa, but can be observed at ~41-55 kDa due to high glycosylation.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HepG2 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Schober et al.
Nature medicine, 20(4), 368-376 (2014-03-04)
Atherosclerosis, a hyperlipidemia-induced chronic inflammatory process of the arterial wall, develops preferentially at sites where disturbed laminar flow compromises endothelial cell (EC) function. Here we show that endothelial miR-126-5p maintains a proliferative reserve in ECs through suppression of the Notch1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service