Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA001048

Sigma-Aldrich

Anti-BMX antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Bone marrow tyrosine kinase gene in chromosome X protein antibody produced in rabbit, Anti-Cytoplasmic tyrosine-protein kinase BMX antibody produced in rabbit, Anti-ETK antibody produced in rabbit, Anti-Epithelial and endothelial tyrosine kinase antibody produced in rabbit, Anti-NTK38 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

KFLCCQQSCKAAPGCTLWEAYANLHTAVNEEKHRVPTFPDRVLKIPRAVPVLKMDAPSSSTTLAQYDNESKKNYGSQPPSSSTSLAQYDSNSKKIYGSQPNFNMQYIPREDFPDWWQVR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BMX(660)

General description

Cytoplasmic tyrosine-protein kinase BMX is an enzyme encoded by the BMX gene in humans and is a member of the TEC kinase family. It is expressed in hematopoietic cells of the myeloid lineage, where it participates in the immune response.

Immunogen

Cytoplasmic tyrosine-protein kinase BMX recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The tyrosine-protein kinase BMX is involved in the response to ischemia and pressure overload in the endocardium and the cardiac endothelium. It encodes a novel nonreceptor tyrosine kinase, which may play a role in the growth and differentiation of hematopoietic cells. It may act as a potential predictor of clinical symptoms following mTBI (mild traumatic brain injury). The overexpression of this gene is associated with malignancy (expressed in several types of cancers) and disease progression of RCC (renal cell carcinoma). It may act as a potential therapeutic target for RCC. It is essential for the activation of the MAP (mitogen activated protein) kinase and NFκB pathways and acts at the level of the essential TAK1/TAB (transforming growth factor beta-activated kinase 1) complex.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74136

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Jia Wang et al.
BioMed research international, 2014, 293687-293687 (2014-05-27)
Mild traumatic brain injury (mTBI) is one of the most common neurological disorders. Most patients diagnosed with mTBI could fully recover, but 15% of patients suffer from persistent symptoms. In recent studies, genetic factors were found to be associated with
Jintao Zhuang et al.
Journal of experimental & clinical cancer research : CR, 33, 25-25 (2014-03-13)
Expression of the non-receptor tyrosine kinase ETK/BMX has been reported in several solid tumors, but the underlying molecular mechanisms and its clinical significance in renal cell carcinoma (RCC) remain to be elucidated. ETK expression in 90 human RCC and 30
Bruno Cenni et al.
International reviews of immunology, 31(2), 166-173 (2012-03-28)
Bone marrow kinase on chromosome X (BMX) is a cytosolic tyrosine kinase and a member of the TEC kinase family. BMX is expressed in hematopoietic cells of the myeloid lineage where it participates in the immune response. It is also
L Tamagnone et al.
Oncogene, 9(12), 3683-3688 (1994-12-01)
The Bmx sequence was identified and cloned during our search for novel tyrosine kinase genes expressed in human bone marrow cells. Bmx cDNA comprises a long open reading frame of 675 amino acids, containing one SH3, one SH2 and one

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service