Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA001761

Sigma-Aldrich

Anti-SEMA3G antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Sem2 antibody produced in rabbit, Anti-Semaphorin-3G precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

VRWLLQRPGDEGPDQVKTDERVLHTERGLLFRRLSRFDAGTYTCTTLEHGFSQTVVRLALVVIVASQLDNLFPPEPKPEEPPARGGLASTPPKAWYKDILQLIG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SEMA3G(56920)

Immunogen

Semaphorin-3G precursor recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

SEMA3G (semaphorin 3G) belongs to the large, evolutionarily conserved family of signaling molecules. Class 3 semaphorins is sub classified into six types named as sema3A to sema3F. SEMA3G acts as a novel peroxisome proliferator-activated receptor γ (PPAR-γ) regulated gene, which is centrally associated with the endothelial cell migration. It is a downstream effector of PPAR-γ. SEMA3G possesses anti-migration and anti-invasion ability.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85018

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weiwei Liu et al.
Journal of cellular biochemistry, 116(4), 514-523 (2014-10-23)
In addition to regulating lipid and glucose metabolism, the nuclear receptor PPAR-γ has emerged as a potentially relevant player in regulating endothelial cell function. Despite the identification of numerous PPAR-γ targets involved in vascular development, the targets downstream of PPAR-γ
Dominique Leitner et al.
Acta neuropathologica, 148(1), 9-9 (2024-07-23)
Cerebral amyloid angiopathy (CAA) is characterized by amyloid beta (Aβ) deposition in cerebrovasculature. It is prevalent with aging and Alzheimer's disease (AD), associated with intracerebral hemorrhage, and contributes to cognitive deficits. To better understand molecular mechanisms, CAA(+) and CAA(-) vessels
Ryoichi Ishibashi et al.
Scientific reports, 6, 25955-25955 (2016-05-18)
Kidney diseases including diabetic nephropathy have become huge medical problems, although its precise mechanisms are still far from understood. In order to increase our knowledge about the patho-physiology of kidney, we have previously identified >300 kidney glomerulus-enriched transcripts through large-scale
Craig B Stevens et al.
Gene expression patterns : GEP, 5(5), 647-653 (2005-06-09)
The semaphorins are a large, evolutionarily conserved family of signaling molecules with broad functions during development. The class 3 semaphorins are a subclass of secreted semaphorins found in vertebrates. There have been six class 3 semaphorins identified to date (sema3A
Xiuping Zhou et al.
Oncology reports, 28(1), 269-275 (2012-05-09)
Glioblastoma multiforme is the most aggressive type of brain tumor with a strong ability to invade and migrate into surrounding normal brain tissues, leading to high tumor recurrence and mortality. Most of class-3 semaphorins, especially SEMA3A, SEMA3B and SEMA3F, have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service