Skip to Content
MilliporeSigma
All Photos(1)

Documents

H6136

SAFC

Hanks′ Balanced Salts

Without sodium bicarbonate, powder, suitable for cell culture

Pharma Manufacturing

Synonym(s):

Hanks′ Balanced Salt Solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Balanced Salt solutions (BSS) are essential in cell culture as they serve as irrigating, transporting, and diluting agents to maintain the osmotic balance between intra-and extracellular spaces. They provide cells with water, inorganic ions, and energy for normal cell metabolism. BSS also provides a buffering system to maintain the medium within the optimum pH range (7.2-7.6).

Application

Hanks′ Balanced Salt has been used:

  • in the preparation of stock solution for culturing mice primary hepatocytes
  • to measure the mineralized nodule formation in human osteoblast cultures
  • in the preparation of embryo buffer to collect zebrafish eggs

Quantity

Formulated to contain 9.8 grams of powder per liter of medium.

Reconstitution

Supplement with 0.35 g/L sodium bicarbonate.

Other Notes

The pH is adjusted after addition of sodium bicarbonate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Behavioral profiling of zebrafish embryos exposed to a panel of 60 water-soluble compounds
Shaukat A, et al.
Behavioural Brain Research, 228, 272-283 (2012)
Osteoblast response to porous titanium and biomimetic surface: In vitro analysis
Renata F, et al.
Materials Science and Engineering, C, 52, 194-203 (2015)
Anish Deshpande et al.
Biomolecules, 10(3) (2020-03-25)
Alpha7 nicotinic acetylcholine receptors (α7nAChRs) are interesting not only because of their physiological effects, but because this receptor requires chaperones to traffic to cell surfaces (measured by alpha-bungarotoxin [αBGT] binding). While knockout (KO) animals and antibodies that react across species
Tom A P Driedonks et al.
Journal of extracellular vesicles, 9(1), 1764213-1764213 (2020-09-19)
Major efforts are made to characterize the presence of microRNA (miRNA) and messenger RNA in blood plasma to discover novel disease-associated biomarkers. MiRNAs in plasma are associated to several types of macromolecular structures, including extracellular vesicles (EV), lipoprotein particles (LPP)
Ming-Fo Hsu et al.
Redox biology, 36, 101658-101658 (2020-08-10)
Alcoholic liver disease (ALD) is a major health problem and a significant cause of liver-related death. Currently, the mainstay for ALD therapy is alcohol abstinence highlighting the need to develop pharmacotherapeutic approaches. Protein-tyrosine phosphatase 1B (PTP1B) is an established regulator

Protocols

Powdered media and salt mixtures are extremely hygroscopic and should be protected from atmospheric moisture.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service