Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H5529

Sigma-Aldrich

Heregulin-α, EGF Domain human

recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥97% (SDS-PAGE)

Synonym(s):

HRG-α

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

10-50 ng/mL

mol wt

protein 8 kDa (reducing conditions)
protein ~7 kDa (predicted)

packaging

pkg of 50 μg

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... NRG1(3084)

Biochem/physiol Actions

Heregulin (HRG) is the human homologue to the neu differentiation factor (NDF) in rat. The heregulin family members contain one EGF-like motif and an IgD-like motif in the extracellular domain. They bind to ErbB-2, ErbB-3, and ErbB-4 (receptors closely related to EGFR). HRG-α and HRG-β isoforms differ slightly in the EGF domain due to alternate splicing. HRG-β isoforms are further subdivided into β1, β2, and β3 isoforms, which show identical binding and activation characteristics. Both α and β HRG isoforms bind to ErbB-3 and ErbB-4 homodimers, but not directly to ErbB-2. HRG-α binding to ErbB-3 and ErbB-4 is reported to be approximately 100-fold weaker than that of HRG-β. When ErbB-2 is combined into a heterodimer with ErbB-3 or ErbB-4, the binding affinities of both α and β isoforms are substantially improved. HRGs are mitogenic for Schwann cells in culture and weakly to moderately mitogenic for a variety of epithelial cells, including mammary, ovarian, lung, and gastric cells. HRGs inhibit proliferation and induce differentation in some tumor cell lines, such as certain mammary tumor cells, which are arrested at the G2M phase. HRGs also induce expression of acetylcholine receptors and possibly other molecules in muscle cells at newly formed neuromuscular synapses, suggesting they may play a role in neuromuscular synapse maturation and maintenance.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline, pH 7.4, containing 50 μg bovine serum albumin per 1 μg as a carrier protein.

Analysis Note

The bioactivity is tested in culture using a proliferation assay with the human cell line MCF-7.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Take your partners, please--signal diversification by the erbB family of receptor tyrosine kinases.
R J Daly
Growth factors (Chur, Switzerland), 16(4), 255-263 (1999-07-31)
S Y Baek et al.
Developmental neuroscience, 20(6), 512-517 (1998-12-22)
Neu differentiation factor (NDF), a 44-kD polypeptide, is a member of the neuregulin family which also includes glial growth factor, heregulin and acetylcholine-receptor-inducing activity. Previous studies have demonstrated that NDF/glial growth factor/heregulin/acetylcholine-receptor-including activity are products of neurons and mediate proliferation
Z Aguilar et al.
Oncogene, 18(44), 6050-6062 (1999-11-11)
The heregulins are a family of ligands with ability to induce phosphorylation of the p185HER-2/neu receptor. Various investigators have reported a variety of responses of mouse and human breast and ovarian cells to this family of ligands including growth stimulation
J T Jones et al.
FEBS letters, 447(2-3), 227-231 (1999-04-24)
ErbB receptor activation is a complex process and is dependent upon the type and number of receptors expressed on a given cell. Previous studies with defined combinations of ErbB receptors expressed in mammalian cells have helped elucidate specific biological responses
Neu differentiation factor (NDF) and the neuregulin (NRG) family.
Yarden, Y., Wen, D. et al.
The Cytokine Handbook, 146-146 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service