Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H2042

Sigma-Aldrich

Histone H2A human

Synonym(s):

Histone, Human

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.25

biological source

human

Quality Level

recombinant

expressed in E. coli

form

liquid

concentration

1 mg/mL

UniProt accession no.

application(s)

cell analysis

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... H2AC20(8338)

Looking for similar products? Visit Product Comparison Guide

General description

Histones are highly basic chromatin proteins. They tightly pack and organize eukaryotic DNA to form nucleosomes. A nucleosome comprises an octameric core histone made of a tetramer of histones H3 and H4 with two dimers of histones H2A and H2B. The 147 DNA base pairs are wrapped around the histone complex. H2A consists of a C-terminal tail extending out of the nucleosome core particle.

Application

Histone H2A human has been used as a neutrophil extracellular trap (NET) to study its effects on the membrane integrity and viability of macrophages (MFS) and dendritic cells (DCS). It has also been used in far-western blot analysis for protein identification.

Biochem/physiol Actions

Histones modulate the accessibility of DNA during transcription, DNA replication, DNA repair, and recombination. N and C terminal tails undergo post-translational modifications (PTMs) altering DNA wrapping and chromatin regulation.

Physical form

Supplied in 20 mM Na3PO4, pH 7.0, 0.3 M NaCl and 1 mM EDTA

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Khoa N Pham et al.
The journal of physical chemistry. B, 125(19), 4977-4986 (2021-05-12)
Histones are highly basic chromatin proteins that tightly package and order eukaryotic DNA into nucleosomes. While the atomic structure of the nucleosomes has been determined, the three-dimensional structure of DNA-free histones remains unresolved. Here, we combine tandem nonlinear and linear
Apurva Bhardwaj et al.
International journal of molecular sciences, 22(1) (2021-01-07)
Insulin-like growth factor (IGF) binding protein-3 (IGFBP-3) is an essential protein that regulates cellular processes such as cell proliferation, apoptosis, and differentiation. It is known to bind with several proteins to carry out various cellular functions. In this study, we
Luis Donis-Maturano et al.
SpringerPlus, 4, 161-161 (2015-04-18)
Neutrophils are one the earliest, crucial innate defenses against innumerable pathogens. Their main microbicidal activities include phagocytosis and degranulation, with many pharmacologically active molecules contributing to inflammation. Recently, a novel antimicrobial mechanism was discovered; the Neutrophil Extracelullar Traps (NETs) formed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service