Skip to Content
MilliporeSigma

H2167

Histone H2B human

Synonym(s):

Histone, Human

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.25
UNSPSC Code:
12352202
Form:
liquid
Biological source:
human
Recombinant:
expressed in E. coli
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in E. coli

form

liquid

concentration

1 mg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... H2BC1(255626)

General description

Histones are highly basic chromatin proteins. They tightly organize eukaryotic DNA into nucleosomes. A nucleosome exists as an octameric core histone comprising a tetramer of histones H3 and H4 with two dimers of histones H2A and H2B, wherein the 147 DNA base pairs are wrapped around the histone complex.

Application

Histone H2B human has been used in far western blot analysis.

Biochem/physiol Actions

Histones are known to regulate the accessibility of DNA during transcription, DNA replication, DNA repair, and recombination. Extrachromosomal histone H2B functions as a cytosolic sensor and detects the double-stranded DNA (dsDNA) fragments produced from infectious agents or damaged cells. This stimulates innate and acquired immune responses in different cell types. Histone H2B moves in and out of the nucleosome more quickly than other core histones.


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Carc. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library