Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE80-6483-37

Sample Grinding Kit

Cytiva 80-6483-37, sufficient for 50 preparations

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
NACRES:
NA.32

usage

sufficient for 50 preparations

feature

wetted part: no

manufacturer/tradename

Cytiva 80-6483-37

packaging

pkg of 50 samples

storage condition

room temperature

Related Categories

General description

Sample grinding kit is ready to use for disrupting small tissue and cell samples for protein extraction.

Application

Sample Grinding Kit is ready to use for disrupting small tissue and cell samples for Protein extraction.

The kit consists of 1.5 mL microcentrifuge tubes, each containing a small quantity of abrasive grinding resin suspended in water, and disposable pestles for grinding. The tube is first centrifuged to pellet the resin and water is removed. Then extraction solution of choice and the sample are added to the tube, and the pestle is used to grind the sample. After centrifugation, cellular debris and grinding resin pellet firmly in the bottom of the conical tube, and the supernatant is easily pipetted out.

Features and Benefits

  • Disrupts small tissue and cell samples for Protein extraction.
  • Uses 1.5 mL microcentrifuge tubes, grinding resin and disposable pestles.
  • Process up to 100 mg of sample per tube in about 10 min.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Sample Grinding Kit from Cytiva is designed to disrupt cell or tissue samples for protein extraction.

Sample Grinding Kit from Cytiva is designed to disrupt cell or tissue samples for protein extraction.

Sample Grinding Kit from Cytiva is designed to disrupt cell or tissue samples for protein extraction.

Sample Grinding Kit from Cytiva is designed to disrupt cell or tissue samples for protein extraction.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service