Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

Z742617

EZ-Grind 20 Pestle & 20 Tubes with Resin

Synonym(s):

DNA extraction equipment, RNA extraction, homogenization system, protein extraction

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.74

packaging

pkg of 20 ea

manufacturer/tradename

G-BioSciences 786-139

General description

EZ-Grind has been specifically developed for effectively grinding small biological samples for proteins and DNA extraction. The resin supplied in the tube contains neutral, high tensile and abrasive micro-particles that effectively disrupt the tissue or cell samples and help in the extraction of all soluble proteins in the extraction buffer. Proteins and nucleic acids do not bind to the resin that can be used effectively grind most animal/plant tissues and cells. Unlike sand or other grinding materials, EZ-Grind is very convenient to work with and does not damage high molecular mass proteins. We routinely use it for extraction of proteins, nucleic acids from different biological samples.

Legal Information

EZ-Grind is a trademark of Geno Technology, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergey Iordanskiy et al.
Retrovirology, 3, 4-4 (2006-01-18)
The early events of the HIV-1 life cycle include entry of the viral core into target cell, assembly of the reverse transcription complex (RTCs) performing reverse transcription, its transformation into integration-competent complexes called pre-integration complexes (PICs), trafficking of complexes into
Timothy S Kern et al.
Diabetes, 56(2), 373-379 (2007-01-30)
Pharmacologic treatment of diabetic retinopathy via eyedrops could have advantages but has not been successful to date. We explored the effect of topical Nepafenac, an anti-inflammatory drug known to reach the retina when administered via eyedrops, on the development of
Sergey Iordanskiy et al.
Retrovirology, 7, 85-85 (2010-10-14)
The impact of the products of the pol gene, specifically, reverse transcriptase (RT) on HIV-1 replication, evolution, and acquisition of drug resistance has been thoroughly characterized for subtype B. For subtype C, which accounts of almost 60% of HIV cases
Sergey N Iordanskiy et al.
Methods in molecular biology (Clifton, N.J.), 485, 121-134 (2008-11-21)
Molecular details and temporal organization of the early (preintegration) phase of HIV life cycle remain among the least investigated and most controversial problems in the biology of HIV. To accomplish reverse transcription and intracellular transport of the viral genetic material

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service