Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE29219757

Superdex® 30 Increase

10/300 GL, Cytiva

Synonym(s):

Size exclusion resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
NB.21

feature

10/300 GL

manufacturer/tradename

29219757
Cytiva

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Superdex 30 Increase SEC columns are for high resolution, small-scale purification and analysis of small biomolecules with Mr ~ 100 to ~ 7000, such as peptides


New generation of size exclusion chromatography columns


Superdex 30 Increase is part of GE’s new generation of agarose-based SEC columns called “Increase”, which offer higher resolution purification in shorter runtimes compared with their predecessors.

Features and Benefits

  • Improved performance – improved resolution and runtime compared to predecessor Superdex Peptide
  • Tolerance to repeated harsh cleaning procedures at high pH ensures long column lifetime and minimizes carryover. purity and analysis results
  • Very high resolution – small bead size and narrow particle size distribution provide high resolution, for high protein purity.
  • High flow rate – due to rigid beads giving excellent pressure/flow properties.

Legal Information

Superdex is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service