Skip to Content
MilliporeSigma
All Photos(1)

Documents

GE28-4038-41

Gel Filtration Cal Kit Low Molecular Weight

Cytiva 28-4038-41

Synonym(s):

Gel filtration calibration

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
NA.25

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 28-4038-41

storage temp.

2-8°C

General description

The Low Molecular Weight (LMW) Kit contains five proteins with Mr in the range 6500 to 75 000 and Blue Dextran 2000, and provides simple, reliable calibration of gel filtration columns.

The kits are suitable for calibration of high-resolution gel filtration media and columns such as Superdex, Superose, and Sephacryl.

Features and Benefits

  • Well defined protein standards that show excellent behavior in gel filtration and enable simple, reliable calibration of gel filtration columns.
  • An optimized range of proteins that suits high-resolution media and prepacked columns.
  • Low and high molecular weight ranges from 6500 to 75000 and 43000 to 669000.
  • Each kit contains five proteins that are lyophilized in individual vials.
  • Includes Blue Dextran 2000 to determine the void fraction in the column.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Skyler Wheaton et al.
The Analyst, 140(14), 4799-4803 (2015-03-06)
We trap a set of molecular weight standard globular proteins using a double nanohole optical trap. The root mean squared variation of the trapping laser transmission intensity gives a linear dependence with the molecular weight, showing the potential for analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service