Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

GE28-9782-45

HiScreen, DEAE Fast Flow

Cytiva 28-9782-45

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101511
NACRES:
NA.56

ligand

diethylaminoethyl

description

Ion Exchanger Type (value)

form

prepacked column

crosslinking

cross-linked

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 28-9782-45

storage condition

(20% Ethanol)

parameter

1.5 bar (22 psi) (Over the Packed Bed During Operation)
22 psi

technique(s)

liquid chromatography (LC): suitable

bed size

7.7 mm × 100 mm

bed volume

4.7 mL

column I.D.

7.7 mm

matrix

6% cross-linked agarose

matrix active group

N+(C2H5)2H functional group

particle size

45-165 μm

average diameter

90 μm

cleaning

2-14

working range

2-12

capacity

binding capacity(~ 110 mg HAS/mL resin)

suitability

suitable for bioprocess medium

separation technique

anion exchange

Related Categories

General description

HiScreen DEAE FF columns are prepacked with DEAE Sepharose Fast Flow, a weak anion exchanger, and are part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening.

Application

The small column volume of 4.7 mL and bed height of 10 cm make HiScreen columns excellent tools for method optimization, parameter screening, robustness testing, and convenient scale-up. Process fluid velocities can be applied, since the 10 cm bed height gives enough residence time and the results can then serve as basis for linear process scale-up.

If necessary, two columns can easily be connected in series to give a bed height of 20 cm.

Media prepacked in HiScreen IEX FF columns are also available in other formats and as bulk packs, making it possible to use the same media for development work, pilot studies, and routine production.

Features and Benefits

  • DEAE Sepharose Fast FLow is a weak anion exchanger and IEX Sepharose Fast FLow medium are well-proven, industrial standards for ion exchange chromatography.
  • Prepacked, ready-to-use columns for convenient process development optimization.
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
  • Easily connected in series to achieve 20 cm bed height.
  • Small bed volume gives fast results and minimal sample/buffer consumption.
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

HiScreen is a trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows volatile and non-volatile buffer suggestions for anion and cation exchange chromatography.

This page covers the standard ÄKTAdesign configurations for simple IEX chromatography.

This page covers practical problems that may lead to a non-ideal IEX separation and their solutions.

This page covers detailed aspects of each step in an IEX separation to improve resolution and overall performance.

Protocols

This page covers the use of Sepharose Fast Flow for purification of proteins.

This page shows how to perform column packing and preparation for ion exchange chromatography and chromatafocusing when using Tricorn or XK columns available from Cytiva.

This page clarifies sample preparation, buffer exchange and desalting, removal of lipoproteins, phenol red, and low molecular weight contaminants in Ion exchange chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service