Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0710-01

DEAE-Sepharose CL-6B

Cytiva 17-0710-01, pack of 500 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

ligand

diethylaminoethyl

description

Ion Exchanger Type (value)

feature

autoclavable In 0.1 M NaCl at 121°C for 30 min

packaging

pack of 500 mL

manufacturer/tradename

Cytiva 17-0710-01

matrix

6% cross-linked agarose

particle size

45-165 μm

average diameter

90 μm

cleaning in place

2-14

working range

3-12

capacity

binding capacity

General description

DEAE Sepharose is a weak anion exchange chromatography resin (medium).
Thyroglobulin (MW 669 000) 2 mg/ml, HSA (MW 68 000) 170 mg/ml, alpha-lactalbumin (MW 14 300) 150 mg/ml

Features and Benefits

  • Composed of crosslinked 6% agarose beads with diethylaminoethyl (DEAE) weak anion exchange groups
  • The resin has high chemical stability, allowing well-proven cleaning-in-place (CIP) and sanitization protocols.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service