Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE28-9365-50

GSTPrep Fast Flow 16/10

Cytiva 28-9365-50

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NA.56

ligand

glutathione and 10-carbon linker arm

packaging

pkg of 20 mL

manufacturer/tradename

Cytiva 28-9365-50

parameter

<10 mL/min flow rate
1.5 bar (22 psi) (Over the Packed Bed During Operation)
22 psi

bed size

16 mm × 100 mm

bed volume

20 mL

column I.D.

16 mm

matrix

highly cross-linked 4% agarose

average diameter

90 μm

cleaning

3-12

working range

3-12

capacity

>10 mg/mL binding capacity

General description

GSTPrep FF 16/10 is prepacked, ready-to-use column containing Glutathione Sepharose 4 Fast FLow designed for one-step, scale-up purification of glutathione S-transferase (GST)-tagged proteins.
The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Application

Glutathione Sepharose 4 Fast FLow is for scale-up purification of recombinant GST-tagged Proteins, other glutathione S-transferases, and glutathione-dependent Proteins. Mild elution conditions preserve Protein antigenicity and function.

Features and Benefits

  • Fast and easy one-step, scale-up purification of GST-tagged Proteins.
  • Binds approx. 200 mg recombinant GST/column
  • Mild elution conditions preserving Protein antigenicity and function.
  • Simple operation with a pump or chromatography system, such as ÄKTA® design.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

GSTPrep is a trademark of Cytiva
Sepharose is a trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows how to cleave and purify GST-tagged proteins eluted from GSTrap from Cytiva.

Protocols

This page how to remove GST tags by enzymatic cleavage using Cytiva products.

Affinity chromatography tags like GST fusion proteins facilitate separations in various purification products from Cytiva.

Affinity chromatography tags like GST fusion proteins facilitate separations in various purification products from Cytiva.

Affinity chromatography tags like GST fusion proteins facilitate separations in various purification products from Cytiva.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service