Skip to Content
MilliporeSigma
All Photos(2)

Documents

GE17-5131-02

GSTrap Fast Flow

Cytiva 17-5131-02, pack of 5 × 5 mL

Synonym(s):

GSTrap Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

ligand

glutathione and 10-carbon linker arm

packaging

pack of 5 × 5 mL

manufacturer/tradename

Cytiva 17-5131-02

parameter

flow rate
42 psi

bed size

16 mm × 25 mm

bed volume

5 mL

column I.D.

16 mm

matrix

highly cross-linked 4% agarose

average diameter

90 μm

cleaning

3-12

working range

3-12

capacity

>10 mg/mL binding capacity

Related Categories

General description

GSTrap FF columns are prepacked Glutathione Sepharose Fast Flow columns for fast, convenient, one-step purification of glutathione S-transferase (GST) tagged proteins.

GST*-tagged proteins can be purified directly from pretreated bacterial lysates using GSTrap FF.GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione. The purification process preserves protein antigenicity and function. If desired, cleavage of the protein from GST can be achieved using a site-specific protease whose recognition sequence is located immediately upstream from the multiple cloning site on the pGEX plasmids. GST-tagged proteins can be detected using colorimetric or immunological methods. The medium, Glutathione Sepharose 4 Fast Flow, is also available as lab packages and is an excellent choice for scale-up.The columns can be operated with a syringe, peristaltic pump, or liquid chromatography system such as AKTA design or FPLC System.

* See the section for licensing information in our terms and conditions.
The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Features and Benefits

  • Fast and simple one-step purification of GST-tagged proteins.
  • Mild elution conditions preserving protein antigenicity and function.
  • Easy scale-up by connecting the columns in series.
  • Sepharose 4 Fast Flow provides good flow properties.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

FPLC is a trademark of Cytiva
GSTrap is a trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows troubleshooting strategies for cleavage methods using Cytiva products.

Protocols

Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.

Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.

Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.

Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service