Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE28-9343-88

HiTrap® Capto IEX Selection Kit

Cytiva 28-9343-88, pack of 1 kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101511
NACRES:
NA.56

ligand

quaternary amine

description

Ion Exchanger Type (value)

packaging

pack of 1 kit

manufacturer/tradename

Cytiva 28-9343-88

parameter

42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

highly cross-linked agarose with dextran surface extender

average diameter

90 μm (d50v)

cleaning

2-14

working range

2-12

suitability

suitable for bioprocess medium

General description

HiTrap Capto IEX Selection Kit is designed for screening the optimal Capto ion exchange or Cytiva multimodal chromatography resin prior to scale-up. This kit contains a variety of columns prepacked with anion and cation exchange resins, allowing users to screen for the most appropriate column prior to scale-up. The kit includes five 1 mL HiTrap columns packed with the following resins:
  • Capto Q strong anion exchanger
  • Capto S strong cation exchanger
  • Capto DEAE weak anion exchanger
  • Capto MMC multimodal cation exchanger
  • Capto adhere multimodal anion exchanger

Features and Benefits

  • Fast and easy selection. Five prepacked HiTrap columns, each containing a different IEX resin, including multimodal ion exchangers.
  • Convenient way to screen for the optimal Cytiva IEX resin for a given application.
  • Excellent for optimization studies, buffer composition, pH, flow rate, and other parameters can be optimized with small samples before scale-up.
  • HiTrap column format is designed for use with a syringe, peristaltic pump, or chromatography system such as an ÄKTA system.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol (Q, DEAE), 20% Ethanol, 0.2 M Sodium Acetate (S))

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

Capto is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page describes principles and standard conditions for different purification techniques of histidine-tagged proteins using Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service